Effects of sample charging
Description:
A sample is being bombarded with negatively charged electrons and the sample becomes negatively charged because it is not a good conductive path for the electrons in the beam to get to ground through the sample.
Environment:
- Noran System 6
- Noran System 7
- Pathfinder
Attachment(s):
File | Last Modified |
---|---|
Effects of Sample Charging.pdf | May 19, 2022 |