Thermo Fisher Scientific
Creating a project template from an existing project
Description: 
After spending some time setting up acquisition parameters, selected elements and processing parameters, I want to save them
Environment:
  • Noran System 6
  • Noran System 7
Resolution:
  1. Open the project with all desired parameters set.
  2. From the main menu File, select Create Template from Project.
  3. A save as window will pop up.
  4. Enter a name for the template and click OK.
  5. Use the default settings for the location. You must have administrative privileges.
  • Was this article helpful?