Calibrate the Laser Frequency
Calibrate your laser frequency if a qualification workflow, such as the Nicolet FTIR - Factory Qualification workflow, indicates a failing test. When a test fails, first align the spectrometer, then calibrate the laser frequency and run the test again. If the test still fails, contact your local technical support representative for help.
To calibrate the laser (touchscreen interface)
- On the home screen, select the Diagnostics icon [
] to open the Diagnostics view.
-
Open the Laser Calibration tab and select Calibrate to begin.
When laser calibration is complete, a message indicates whether the laser frequency has been changed and displays the new frequency.
To calibrate the laser (operator interface)
- Navigate to the Administrator area, and select the Diagnostics icon [
] to open the Diagnostics view.
-
Open the Laser Calibration tab and select Calibrate to begin.
When laser calibration is complete, a message indicates whether the laser frequency has been changed and displays the new frequency.
To calibrate the laser (desktop interface)
- Select Acquire Data > Diagnostics > Laser Calibration.
-
In the Laser Calibration dialog box, click Start to calibrate the laser.
When the system is finished calibrating the laser, a message indicates whether the frequency was changed and displays the new frequency.