Thermo Fisher Scientific
Unable to Start PV Test in Insight Pro with Security Suite
Issue
Customer with Insight Pro  is unable to start a PV Test
Environment
Insight Pro with Security Suite
Resolution
First make sure all Security services are running. The users need to have Modify, Read, and Write permissions to the folder C:\ProgramData\Thermo\Insight and all sub-folders. 
Analysis
Cause