Thermo Fisher Scientific
Safety and operating precautions
Prevent fire and explosion by using an active venting system free of spark and ignition sources when using volatile solvents. Protect sample windows from thermal and mechanical shock, avoid strong acids, bases, oxidizing agents, and chlorinated solvents. Read the safety guide before operating the spectrometer.
Prevent fire and explosion by using an active venting system free of spark and ignition sources when using volatile solvents. Protect sample windows from thermal and mechanical shock, avoid strong acids, bases, oxidizing agents, and chlorinated solvents. Read the safety guide before operating the spectrometer.
CAUTION_Flammable.png Prevent fire and explosion. If using volatile solvents, provide an active venting system that is free of spark and other ignition sources and prevents flammable vapors from collecting in the atmosphere surrounding the instrument.
To prolong the life of your system, follow these precautions:
  • Protect sample windows from thermal and mechanical shock
  • Do not leave solvent on the sample windows for longer than 2 minutes.
  • Do not attempt to measure materials that are strong acids (pH less than 4), strong bases (pH greater than 11) or strong oxidizing agents
  • Do not use chlorinated solvents or materials that may produce HCl or HF vapors with or near the system
  • Do not block the vents at the back of the spectrometer
Read the safety guide that came with the spectrometer before operating the spectrometer with the Affirma Lubricant Analysis System installed.
  • Was this article helpful?