Thermo Fisher Scientific
Unpack the FTRN Accessory
To prevent condensation damage, leave the FTRN accessory in its plastic shipping bags for 4 hours after delivery to adjust to room temperature. Save packing materials for future use. After 4 hours, unpack all components, verify the order against the packing list, and remove components from their plastic bags.
To prevent condensation, which can damage internal optics, leave the FTRN accessory and components in their plastic shipping bags for 4 hours after delivery to allow them to slowly adjust to room temperature. Save the packing materials for storing or transporting the accessory.
To unpack the FTRN Accessory
  1. Remove all components from their shipping boxes.
  2. Wait 4 hours and then remove any components from their plastic shipping bags.
  3. Verify the order is complete (compare against packing list).
  • Was this article helpful?