Thermo Fisher Scientific
OMNIC Anywhere - August 2023 Release
This OMNIC Anywhere release includes a new Multi-component Search feature for library subscribers and improvements to Compound Search results. Users can now view Multi-component Search results in overlay and stacked views and receive email and in-app notifications. Additionally, Connect Team Edition users can import new file types, including .spc, .sp, and .jws.
This OMNIC Anywhere release includes the new Multi-component Search feature for users who have purchased a library subscription. Additionally, improvements were made to Compound Search to provide better results.
What’s included:
  • New Multi-component search feature for library collection subscribers.
  • Overlay and stacked views for Multi-component Search results.
  • Multi-component search results email and in-app notifications.
  • Compound Search results improvements.
To run a multi-component search:
  • In the Measurements screen, right-click on any measurement with absorbance y-axis units
  • Select Multi-component search
Screenshot 2023-08-23 at 2.03.56 PM.png
  • Select the number of components and libraries for search
Screenshot 2023-08-23 at 2.07.33 PM.png
  • Click on Run search
  • The search will be performed in the background and you will receive an email and app notification once it's completed.
  • When you receive the notification, click on the link to view your results.
Screenshot 2023-08-23 at 2.12.47 PM.png
  • You can view multiple sets of results by clicking on the left panel...
Screenshot 2023-08-23 at 2.24.51 PM.png
... and you can switch between an overlay and stacked view.
Screenshot 2023-08-23 at 2.26.52 PM.png
Lastly, Connect Team Edition users can now import new types of files to OMNIC Anywhere, including .spc, .sp, .jws, and others. For more information about Connect Team Edition, visit this page.
  • Was this article helpful?