Thermo Fisher Scientific
OMNIC Anywhere - November 2023 Release
This OMNIC Anywhere release includes improvements to the Multi-component Search feature for library subscribers, allowing results to be exported as a PDF. Multi-component search is now compatible with measurements in Transmittance, Reflectance, and other units. Additionally, an issue related to libraries within correlation search has been addressed, along with other minor fixes.
This OMNIC Anywhere release includes improvements to our Multi-component Search feature for users who have purchased a library subscription. Additionally, some minor fixes were implemented.
What’s included:
  • Multi-component search results can now be exported as a PDF.
  • Multi-component search is now compatible with measurements in Transmittance, Reflectance and other units.
  • An issue related to libraries within correlation search has been addressed, along with other minor fixes.
To export Multi-component Search results as a PDF file:
  • Run a correlation search by following the instructions in this article
  • In the results screen, choose your preferred view (Stack or Overlay)
Screenshot 2023-11-22 at 2.52.36 PM.png
  • Click on Download PDF to get a report using your preferred view
Screenshot 2023-11-22 at 2.53.32 PM.png
Screenshot 2023-11-22 at 2.54.42 PM.png
  • Was this article helpful?