Thermo Fisher Scientific
OMNIC Anywhere - February 2023 Release
This OMNIC Anywhere release includes a new visualization tool for identified functional groups, improvements to Spectral View and Support pages, and other minor enhancements and bug fixes. To view functional group ranges, select a measurement, click Identify functional groups, and check the boxes of the groups you want to visualize.
This OMNIC Anywhere release includes improvements for functional groups, Spectral View and our Support section.
What’s included:
  • A new visualization tool for identified functional groups.
  • Improvements for Spectral View and Support pages.
  • Other minor improvements and bug fixes.
To view functional group ranges:
  • Select a measurement and click Identify functional groups.
Screen Shot 2023-02-20 at 3.07.29 PM.png
  • Click on View details.
Screen Shot 2023-02-20 at 3.08.05 PM.png
  • Click on the check-boxes of the functional groups you want to visualize.
Screen Shot 2023-02-20 at 3.09.04 PM.png
  • Was this article helpful?