OMNIC Paradigm Software Release Notes
About This Document
This document contains a revision history of OMNIC Paradigm software, including new features that may not be included in the user guide, resolved issues and known issues.
OMNIC Paradigm 2.8 HF2
-
Build number: 2.8.2.38
-
Release date: September 2025
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
Resolved Issues
-
Using ATR Correction command in workflow on a spectrum with invalid y-axis units would fail and would destroy the spectrum. Now the software will not allow you to run the ATR Correction command on a spectrum with invalid y-axis units.
-
You were unable to gather log files or export sessions when bundling very large files. If you attempted to gather log files or export sessions when the files were very large, the software would fail to bundle the files. This is now fixed, and the software can bundle these large files.
-
The software attempted to collect background measurements when the final format was interferogram or single beam. Now the software correctly skips this step when the final format is interferogram or single beam.
-
When using Rapid Scan, you could not use Start from External Trigger. The software now allows you to use Start from External Trigger when using Rapid Scan.
-
Measurements would fail when using Rapid Scan with a velocity of less than 1 cm/s. Rapid scans with a velocity less than 1 cm/s would fail. This issue caused measurements to fail due to "too many failed scans." The fix ensures that Rapid Scan can only be used with supported velocities.
-
During measurements, the displayed number of scans completed during a measurement temporarily dropped to 0. This was a display issue only and did not affect the measurement. The software now correctly displays the number of scans completed during a measurement.
-
For some systems, the System Status dialog showed a blank screen after the initial setup. This is now fixed, and the System Status dialog shows the correct information.
-
The software did not save the X-axis format after changing it. When reopening the measurement, the X-axis format may have been incorrect or the data may not have been displayed correctly. The software now saves the X-axis format and displays it correctly when reopening the measurement.
-
Background settings were not saved with your measurement’s settings. If you named and saved your measurement settings, the background settings were not saved with them. This is now fixed, and the background settings are saved correctly.
-
The template report for QCheck did not show the sample spectrum. The report now correctly shows the sample spectrum.
Time Series Measurements
-
Library searches in time series measurements showed results incorrectly if the main profile was using units other than minutes. This is now fixed, and the search results are shown correctly regardless of the units used in the main profile.
Workflows
-
The Nicolet FTIR - PV Workflow would not run with APEX spectrometers using a ZnSe beamsplitter. This is now fixed, and the workflow runs correctly with APEX spectrometers using a ZnSe beamsplitter.
-
When using the Nicolet iS50 ATR Module, the PHEUR Qualification workflow would incorrectly check the main sample compartment. The workflow now correctly ignores the main sample compartment. The workflow tests only the ATR beam path.
-
After running the Factory Qualification workflow with the FT-Raman module, the blue laser light would not turn off. The blue laser light now turns off after running the Factory Qualification workflow with the FT-Raman module.
-
The history events in spectrum information did not always show if QCheck was run in a workflow. The history events now show if QCheck was run in a workflow.
-
When using iTX accessory, the "Nicolet FTIR - PV Test" workflow would not recognize when you removed the accessory and could not continue. The workflow now correctly detects when you remove the iTX accessory and continues without failing.
-
When using the RaptIR microscope, the stage would not initialize correctly during some workflows. These workflows now correctly initialize the stage.
FT-Raman Module
-
When working in a FT-Raman session, selecting to turn on the laser light would not reliably turn on the laser. The software now reliably turns on the laser light when you select to turn it on in a FT-Raman session.
-
The aperture was set too small when using the FT-Raman module. This is resolved, and the aperture is now the correct size.
Microscopy
-
In a microscopy session, the background list in the region queue was empty after measuring background points. Because of this, you cannot assign specific background points to regions in the region queue.
-
Points measured in a session were labeled inconsistently with Area and lines. Now, all types of regions are labeled consistently.
-
When Prompt for Session Name was selected, the prompt was skipped when starting a session from the toolbar. Now users are correctly prompted for a session name when starting a session from the toolbar.
-
When switching from the Raman module to RaptIR microscope, a backend service would crash. This is now fixed, and the backend service does not crash when switching from the Raman module to the RaptIR microscope.
-
The icon showing the current microscope objective in the camera View did not update in some situations. This is now fixed and the icon updates correctly.
-
When viewing particle analysis results, the displayed spectrum was not always the exact compound selected in the Spectra Matches table. Now, the displayed spectrum is the same compound listed in the Spectra Matches table.
Security Administration
-
In some situations, it was possible for users to run workflows even when the Security Administration software should have prevented them from doing so. Users can no longer run workflows if they do not have permission to run them.
OMNIC Paradigm 2.8 HF1
-
Build number: 2.8.1.31
-
Release date: April 2025
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
Resolved Issues
Workflows Failing
Resolved an issue where running many workflows could cause them to fail.
Failure in Collecting Very Large Maps in Microscopy
Fixed a timeout issue that occurred while the stage was moving during the collection of very large maps.
X- and Y-Axis Values Swapped in TSV File Imports
Corrected the issue where TSV files were imported with the X- and Y-axis values swapped.
System Crash When Saving or Exporting Custom Mosaic Images
Fixed a crash that occurred when saving or exporting custom mosaic images.
Incorrect Photometric Linearity Test Results in NIR System workflows
Resolved incorrect or unusual results in photometric linearity tests for the NIR system when running workflows in languages other than English.
Inability to Import Certain Legacy Map Files
The software now displays an error message when attempting to import unsupported legacy map files.
Artifacts in Time Series Measurements Using Rapid Scan
Fixed the issue where using Rapid Scan in Time Series measurements caused artifacts in the data. Note that measurements collected with Rapid Scan in version 2.8 may still contain these artifacts.
Incorrect X-Values in Time Series TSV File Exports
Resolved an issue where exporting a Time Series measurement as a TSV file saved the x-values as all zeroes.
Incorrect User Name and Spectrum Title in Workflow Reports
Reports created in a workflow now correctly display the user name and do not include unwanted information in the spectrum title.
Error Displaying Annotations After Scaling Spectra
Fixed an error that occurred when displaying annotations, such as peak locations, after scaling spectra.
OMNIC Paradigm 2.8
-
Build number: 2.8.0.793
-
Release date: January 2025
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
New Features
General
New sampling information options. Control how spectra are displayed, including more sampling information, by using the sampling information tab in the Options dialog. You can also access these options from a Spectra tab to control how information is shown on only the data on that tab.
You can now retrieve interferograms from a measurement for reprocessing or other purposes. In a Spectra tab, select a measurement and then go to the Process menu and choose Retrieve interferograms. The interferograms are added to the Spectra tab for further analysis or processing.
Reprocess interferograms and single beam spectra using the Reprocess tool in the Process menu.
Print your data exactly as its displayed by using the CTRL+P keyboard shortcut. In a spectra, time series, or session tab, press CTRL+P to quickly print your display. This can be faster than creating a report, but reports provide richer controls over which data is included.
With the new Split tool, you can export spectra from series and session measurements. For example, using the Region Queue in a session analysis tab, you can use Split to export all the spectra from a measured region. The exported spectra can be saved to files or added directly to the dashboard.
Microscopy
-
Use the new virtual aperture in the Camera view with a microscope to easily visualize, resize, and rotate the microscope aperture.
-
Right-click a mosaic image to export, copy, or print the mosaic image.
-
Apply the Kramers-Kronig correction to all data in a session.
Workflows
-
The QCheck tile now allows loading a reference directly from the database without needing a Load Spectrum tile.
Enhancements
General
-
You can now rename measurements from the dashboard when you select More and view measurement details.
-
Reports now have better support for landscape view, allowing a wider area for spectra.
-
The history tab in the spectrum information pane now shows when spectra were imported from a file.
-
Create a multi-spectrum report from a search result.
-
Measurement settings now update immediately when changing the sampling location or accessory.
Microscopy
-
Session ending process improved. The software now clarifies that the session will be saved and that you should remove the sample before proceeding.
-
Camera view maintains size and position when moving to the acquisition tab.
-
From the dashboard, you can now export multiple spectra as TSV files. Previously, this file type was not an option.
-
Selection checkboxes in the Region Queue have been clarified.
-
The spectral preview pane in the Analysis tab now shows which Region the spectrum belongs to and the XY location.
-
Send a background measurement to the dashboard as a standalone measurement. Before, you could only send sample measurements.
Workflows
-
Qualification workflows now require only one signature for the entire workflow.
-
Added more qualification options for the Apex system with ZeSe.
-
Modified RaptIR qualification workflows support systems using XT-KBr and ZnSe beamsplitters.
-
Workflows use less memory when using a Search tile.
Security
-
Added security controls for performance tests in the Diagnostics menu.
Resolved Issues
General
-
Measurements added from the dashboard to an open tab no longer hide other spectra.
-
Resolved issues caused by adding or editing comments or custom fields in the spectrum information.
-
Fixed Multi Range Processing where it was not stitching two spectra together properly.
-
Improved software performance on very high-resolution displays.
-
Corrected the issue with the NIR Integrating Sphere sample location setting the attenuation wheel to None.
-
Minor formatting issues in Search reports have been corrected.
-
Fixed printing issue in the History tab where information after the first page was cut off.
-
Resolved peak annotation issues in Peak reports where labels were cut off or overlapping.
Microscopy
-
Corrected compatibility report errors between background and measurement settings on the iS20 spectrometer with RaptIR microscope.
Time Series Measurements
-
Fixed color mismatch issue between the spectra display and the right panel during a component search.
-
Corrected profile curve display issue for languages using a comma as the decimal separator.
-
Fixed inconsistency in automatic baseline correction results between Time Series and normal spectra tab.
Workflows and Qualification
-
Fixed NIR qualification workflows to correctly show a preview image.
-
Resolved printing issue with unnecessary whitespace in factory qualification workflow reports.
-
Fixed error when using a quant result as a Pass/Fail check in non-English workflows.
Security
-
Resolved issue that was allowing access to administrator options in Operator mode without a password.
-
Signatures are now required when duplicating, deleting, renaming, exporting, or editing a workflow.
-
Fixed audit log issue where only one spectra signature was recorded when signing multiple spectra.
-
Corrected audit log to record events showing measurement signatures in Spectra tab.
Known Issues
-
Exporting multiple files does not work if the folder has a '.' character in it. For example, if the file path includes a folder named "firstName.lastName", it will get changed to "firstNameLastName" and then not recognize it as a valid path, so Save is disabled.
-
Viewing series data in 3D view may fail. If this occurs, close the 3D view and then reopen it.
-
When measuring multiple single points in a session, some points may be missed. These may appear as a blank or empty spectrum in the sample list in the Acquisition tab.
OMNIC Paradigm 2.7
-
Release date: June 2024
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
New Features
Advanced spectral math in workflows
You can now perform advanced spectral math in a workflow with the new Advanced Spectral Math tile. Before, you carry out simple operations with the Processing tile. With the Advanced Spectral Math tile, you can create custom equations and use more advanced operations. The resulting spectrum is available in other tiles, such as reports and template reports.
Security features
Managed workflows and the secure data folder
With Security Suite software, this new version of OMNIC Paradigm software introduces managed workflows. Managed workflows build on previous security features to offer more control over workflows. Previously, administrators could use access controls and security policies to control which users could edit, open, or run workflows and more. With managed workflows, administrators can also enforce their business’s standard operating procedures by controlling a workflow’s state. Using secure data folders, the administrator can control whether workflows are in development, approved for use, or archived and can control who can use them in each state.
For more on enabling and using this new feature, see the OMNIC Paradigm software user guides.
Enhancements and Resolved Issues
Security
The required signature order was not enforced when signing reports
If you were enforcing a signature order, the order should have been enforced on reports as well as measurements. Previously, when you signed reports, the software did not always check the signature order. This is now fixed.
If you signed multiple measurements at once, some were not locked
If you selected a group of measurements and signed them all at once, not all of them would have been locked when they should have been. Now when signing a group, all those that should be locked are locked.
We revised the report for the accessory performance test workflow
The previous version of the report shown during an accessory performance test workflow was confusing and showed potentially misleading values, even if the test was passing and the values were correct. We have revised the report to clarify the units. This issue did not cause any false passes or failures.
If you deleted an annotation, the software may have deleted the wrong annotation.
The software incorrectly deleted the most recent annotation rather than the selected annotation. This is fixed.
When using the print button on a report tile in a workflow, the report would not show signatures
If you used the print button when viewing a report in a workflow, the report would not show the signature. Reports created elsewhere correctly showed the signatures, but these did not. This is now fixed and the reports show signatures when they are signed.
Microscopy
Microscopy sessions would not save when closing if they did not have a 4x mosaic image
This is now fixed and sessions save when closing if any mosaic is present, including if there is only a 15x mosaic but not 4x mosaic.
When you opened the Region Queue, it appeared mostly off-screen.
This is corrected so that it is now completely visible when you first open it.
There were problems when viewing or saving very large session files with large mosaic images
Previously, the software had difficulty opening or saving very large session files with large mosaic images. This has been improved and the software is better able to handle these large datasets.
Now you can resize the camera view
In older versions of software, you could not change the size of the camera view.
Imported sessions now show up without having to refresh the list
Before, if you imported a session, it would not always appear in the list of sessions on the dashboard. You needed to refresh the list before it was shown. Now the list is updated automatically when the session is imported.
You could not recover the camera feed if it was disconnected while in use
For example, if the USB cable was disconnected during use, the software could not reconnect to the camera after plugging it back in. Now the software can recover the camera feed.
An issue may have caused duplicate images in mosaic images
For example, if the nose was jostled while collecting a 4x mosaic image, the system may have added duplicate images to the mosaic rather than new images. This is fixed and the system is now able to recover when there is an issue and prevent duplicate images.
Spectra sent from a session file were missing metadata
If you sent spectra from a session file, the sent spectrum were missing metadata or would show incorrect metadata. For example, the detector type may have shown ‘unknown’ rather than the correct detector type.
The dashboard did not show a preview image for sessions that did not have a 4x mosaic.
If your microscopy session did not have a 4x mosaic image saved, the preview on the dashboard did not show any preview image. Now the preview shows the 15x mosaic if there is no 4x mosaic.
The RaptIR microscope stage would not lower when the ATR settings menu was open.
If you had the ATR settings menu open, the software would prompt you to install the ATR but the stage would not drop to make room for the accessory. Now the stage correctly moves out to the way.
Microscopy sessions would close without confirmation if you changed the selected sample compartment
If you changed the selected sample compartment while a microscopy session was open, the session would close without prompting you to confirm that you wanted to end the session. Now when you change sample compartments, the software confirms that you want to close the session first.
For the Raman module
The camera view is now available from the dashboard when using the Raman module
You can now open the camera view from the toolbar while viewing the dashboard. Now you can view your sample while reviewing your initial measurement settings.
Control the step size when using the Raman module.
Open the Raman module controls in the FT-Raman view to control the step size for your data collection.
OMNIC Paradigm 2.6
-
Release date: April 2024
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
New Features and Enhancements
See the updated user guide for OMNIC Paradigm software on our knowledge base for more information about using these new features and enhancements.
General Features
New Performance tests
The new performance test for the Nicolet iS50 spectrometer checks your combination of beamsplitter, detector, and sampling location to verify that your instrument setup will measure high-quality data. To use this test, go to the Acquire Data menu and select Diagnostics > Performance Test.
Manual baseline correction
When viewing spectra in the Spectrum tab, you can now correct the baseline manually to ensure that you view the spectrum exactly as you want it. You can still use automatic baseline correction for a quick, accurate correction.
New option for the x-axis format in the spectral view
In the Options dialog, you can now set the x-axis format to split at 2000 cm-1. In this format, the x-axis is split into two regions with a change of scale at 2000 wavenumbers. The scale of the region below 2000 wavenumbers is expanded by a factor of two as compared with the scale of the region above 2000 wavenumbers. Note that this setting only applies to spectra in the Spectrum tab and not in reports or session tabs.
Change the look of the text
Use the new font settings in the Options dialog to customize the look of the text in the spectrum view and other data view. These font choices do not apply to reports.
It is easier to find and organize all your data
Now you can search for workflows and packages by name, just like you were able to before with measurements, reports, and other data. And with the new tag management tools, you can view all the tags you have already used.
Easier to work with tabs
You have more control over tabs and can send data to a tab more easily with the new controls in this version. Right-click the name on a tab to rename it. From the spectrum view, navigate to the spectrum history and right-click to duplicate a measurement and send it to a specific tab.
Measure background after collecting data
View your background settings if you want to measure the background after the sample instead of before the sample. Before, you could only measure the background before measuring the sample or use a previously measured and saved background spectrum. Now you can measure your sample and then the background.
More control over your saved reports
In this release, you can now rename or delete your saved reports, just like saved measurements. If you need to control access to these options, we recommend using our Security Suite software.
The History tab in the Spectrum view now shows more changes to the spectral metadata
If you edit the tags on a spectrum with the Manage Tags tool or add comments or custom fields, the History tab in the Spectrum shows these changes.
Rapid scan available when using a Nicolet Apex spectrometer
If you are using the Nicolet Apex spectrometer, you can enable the Rapid Scan option in Time Series measurements to capture data more quickly. This is useful if your samples change quickly but it will require more storage space to save the full Time Series measurement. Note that for now, this option is available only with the Nicolet Apex spectrometer.
Time Series Measurements
These features and enhancements apply to Time Series measurements.
3D Analysis tools
This version of OMNIC Paradigm software introduces 3D analysis for your Time Series measurements, including waterfall and surface plots. This view provides another way to visualize and analyze your Time Series data.
Note: 3D features should run on any graphics system (board and driver) that provides a complete implementation of OpenGL 4.1. Please note that some features may require OpenGL 4.6. However, boards and driver bugs are not unusual. We recommend using a professional graphics board to benefit from the professional support offered by the vendors (driver bugs fixes, etc.).
Use a previous background measurement for your time series analysis.
Now you can use a background measurement that you measured and saved earlier instead of measuring before collecting sample data. This option is not yet available for measurements with the RaptIR microscope or the Raman module.
Blank a time region in a Time Series measurement
You can blank a region of time in your Time Series measurements.
Split Time Series data into individual measurements
Select a region in your Time Series data and send all the spectra in that range to the dashboard as individual measurements or select a single point to send only one spectrum. From the dashboard, you can quickly export these measurements as files.
Workflows
This version of OMNIC Paradigm software introduces two new workflow tiles and adds more options to familiar tiles.
Read or modify data about your spectrum with the new Spectral Metadata tile
With this new tile, you can read data from the spectrum and use it later as a variable or you can change the data to a new value. For example, you can set the spectrum title to a new custom value or set it equal to an existing variable. Or you can read the data to convert it to a variable that you can use elsewhere in the workflow, such as in a Report tile.
Add to, select from, and iterate through lists with the new List tile
The List tile is useful for modifying a list or reading data from a list. For example, this is useful when paired with Repeat tile. You can use the number of times the loop has repeated to automatically select an element from the list and then perform some processing or analysis on that item. This is a straightforward way to repeat the same processing on every spectrum in a list.
Import a list of spectra with the Load Spectrum tile
Now you can import a list of spectra with the Load Spectrum tile instead of only a single spectrum. Modify or read from the list with the List tile.
Updates to the Measure tile
The Measure tile now has new options, including the ability to match the measurement settings from a SPA, QNT, or EXPX file or a measurement in your database. Select Match Settings and select the source file or measurement to match.
Resolved Issues
Software would crash if you switched to viewing Time Series measurements on the dashboard
For some users, the software would close unexpectedly when trying to view Time Series measurements on the dashboard. This is now fixed so you can safely switch to viewing your saved Time Series measurements.
The results from some search operations were in an incorrect format
When performing a multi-component search or some other library searches, the results may not have matched the final format of your spectrum. For example, if you searched using a spectrum in %T format, the results may have been in absorbance units. The results are fixed to now match the format of the spectrum used in the search.
The Export tile in a workflow now correctly exports SPC files
Previously, when you attempted to export a SPC file with the Export tile in a workflow, the workflow would incorrectly export an SPA file. The tile now works correctly to export SPC files.
Moving a label after peak pick
Peak annotations were incorrect if you moved the label. This is fixed so that the peak values remain correct even after moving labels.
Open spectrum view showing mixed previews for SPG files
When you viewed the preview of an SPG file in the Open dialog, clicking a second SPG file would add the preview to the preview of the previous file. Now the preview shows only the correct SPG file.
Known Issues
The Smart NIR Accessory does not flip the internal mirror during user-created workflows
If you use the Smart NIR Accessory in a workflow, the NIR Background Mirror setting does not correctly move the mirror in or out of the beam path. This only affects user-created workflows. The mirror works correctly in all qualification workflows.
As a workaround, enable Use Sample Shuttle in the Measure tile to move the mirror in and out of the beam path.
Factory-created workflows will sometimes fail to run
Previously, factory-created workflows would sometimes open in the Workflow Editor view instead of running. This no longer happens, but in certain circumstances, the workflow may fail to run. If this happens, the software prompts you to restart the software and run the workflow again.
The Run button in the Workflow Editor and the Camera view in microscopy interfere with one another
Occasionally, these features will interfere with one another and prevent the other from working. For example, if you use the camera view in an FTRaman session, you may not be able to use the Run button in the Workflow Editor afterward. If this happens, the software prompts you to restart the software.
Spectra sent to the dashboard from a Session are missing metadata
If you send spectra to the dashboard from a Session, the spectral metadata may be missing information or showing incorrect information.
Imported sessions not showing in the Sessions list on the dashboard
When you import a session file, the new session is not added to the list of sessions on the dashboard immediately. To see the new session, you must either click Refresh to update the list of sessions or restart the software.
Repeatedly converting the x-axis units can cause errors
If you convert the x-axis units several times between wavenumbers, microns, and nanometers, errors may occur.
OMNIC Paradigm v2.5 HF1
Release Date: October 2023
Supported Operating Systems: Windows 11 64-bit; Windows 10 64-bit
Resolved Issues
Incorrect values when importing spectra data
%T values were 100 times greater than they should be when you imported these files types:
-
Grams (*.spc)
-
JCAMP-DX (*.jdx)
-
Perkin-Elmer (*.sp)
-
Jasco (*.jws)
This is fixed and the software now displays the correct values for %T.
Repeated prompt asking if you trust the connection to a Nicolet Summit or Summit PRO spectrometer
A bug caused you to have to indicate that you trust the connection every time you started the software and connected to a Nicolet Summit or Summit PRO spectrometer. This is fixed, and the software now remembers if you trust the connection.
OMNIC Paradigm v2.5
Release Date: September 2023
Supported Operating Systems: Windows 11 64-bit; Windows 10 64-bit
New Features
General Features
OMNIC Paradigm software now supports more file types
You can now import spectral data from other applications, including the following:
-
Grams (*.spc)
-
JCAMP-DX (*.jdx)
-
Perkin-Elmer (*.sp)
-
Bruker (*.[0-9])
-
Jasco (*.jws)
-
TSV Text (*.tsv)
Enhancements for using OMNIC Paradigm software with Security Suite software
The new version of OMNIC Paradigm software supports a full audit trail for tagging, including tags created in workflows, supports showing multiple digital signatures in reports, and shows the database and security server hostnames in the system status information. We have also added a security policy in Security Administration software to allow you to buffer and save data.
The software now uses HTTPS
OMNIC Paradigm software now uses HTTPS instead of HTTP. This means that the data you send over the network is encrypted for improved security and privacy. This change also means that the first time you connect to a remote instrument, such as a Summit spectrometer on your network, you will be asked if you trust the instrument. This is expected.
OMNIC Paradigm Configuration Utility
The new OMNIC Paradigm Configuration Utility is used to set up and customize your SSL certificate for the software. This utility should be used only by administrators and should use only the default settings unless your organizations policies require a change.
Time Series Measurements
Zoom in on profiles
You can now zoom in on the profile pane for a closer look at the profile. Right-click the profile and select zoom. Then, click and drag to draw a box and click in the box to zoom to that area. Double-click to return to the full profile.
Save interferograms with your time series measurements
You now have the option to save interferograms with the time series data. After setting your analysis type as Time Series, select More to view more settings. Saving interferograms will increase the size of the saved data.
Add comments and custom fields to time series measurements.
Now you can add comments and custom fields to time series data, just like you could with standard measurements previously. To add a comment or a custom field, select Time Series in the measurements pane of the dashboard and then select More to view details.
Import legacy time series measurements from OMNIC software
You can now import legacy Time Series files (files with a .srs file name extension), allowing you to work with your older OMNIC data in OMNIC Paradigm software.
Workflows and Packages
New qualification workflows for the Nicolet iN5 microscope
OMNIC Paradigm software now supports a suite of qualification workflows for the Nicolet iN5 microscope.
Analyze peak areas with the Peak Analysis tile
The Peak Analysis tile now supports the Peak Area and Peak Area Ratio operations.
Microscopy
Support for the Nicolet iN5 Microscope
OMNIC Paradigm software now supports the Nicolet iN5 microscope. See the new Nicolet iN5 Microscope with OMNIC Paradigm software user guide for more information.
Apply atmospheric suppression after collecting data
You can now apply atmospheric suppression to your session data after finishing your measurement. Previously, you had to set this before measuring.
Live spectrum view shows interferogram minimum, maximum, and range values
The live spectrum view now shows the minimum and maximum voltage for the interferogram and the total difference or range.
Better filters and search with security
When finding saved sessions, you can now use security-related filters to find sessions more quickly. Before, you could do this with measurements and time series data but not with sessions.
The Live Spectrum pane shows a saturation warning
The Live Spectrum pane now shows a warning that the detector is saturated when the interferogram voltage is too high.
Use the RaptIR microscope as a right-side emission source
If your microscope is installed on the right-side of your spectrometer, you can now use it as emission source. Select right-side collimated as your source in the software.
The camera view available on the dashboard
We added the Camera View to the toolbar on the dashboard so that you can more easily view your sample before you start your session.
Added Roundness to particle analysis results
Particle analysis now shows a value for roundness in addition to the previous results.
Raman Module: Turn off the laser after a period of inactivity
When using the Raman module, you can now have the laser turn off automatically after a set period of inactivity. This can help protect your sample from accidental damage.
Raman module: The system will automatically cancel
If you open the accessory door or turn the laser off while collecting data, the system will now automatically cancel and will stop collecting.
Resolved Issues
General Features
Improved spectral images in reports
Reports now show a smoother, higher-resolution image of your spectra.
Reports from a Factory Qualification workflow did not always open in the Touchscreen view.
This has been resolved the reports now correctly open when using the Touchscreen view.
Unexpected initial value when calibrating the laser
When calibrating the laser, if the actual laser frequency was outside of the range the software expected, the software would start the calibration at a default value before calibrating. Calibration was working correctly but the screen showed an incorrect starting value. The software now shows the correct, actual starting frequency.
Problem with navigation in the Touchscreen view
When using the Touchscreen view, when doing a Search or QCheck analysis, if you set your Final Format to Single Beam, you may have been unable to navigate to other setup tabs, such as the Summary tab. Single Beam was not a valid option for these types of analysis, so we removed this option and you can no longer encounter this issue.
Missing result in the Quantification and Algorithms Workflow report
The Quantification and Algorithms workflow report was missing the result of the Similarity Match test. The test was still running but the report was not showing the result. This is fixed and the report now shows the Similarity Match result.
Default time range is now All instead of Past Week
By default, the measurements pane now shows you all your measurements, reports, and other saved data rather than showing only data collected in the past week.
You can no longer set your beamsplitter in the software if you do not have an ABX.
It was possible to select a beamsplitter in the software that was not actually installed in the instrument. Now, if you do not have an ABX installed, the beamsplitter is read from the instrument and cannot be incorrectly changed.
Microscopy
Improved file handling when closing a session
Previously it was possible to close the software without saving your session. Now when you close a session, you always have the option to save your data or to cancel and continue working.
Improved the circularity calculation in particle analysis
The previous calculation for circularity when doing particle analysis was incorrect. We have improved the calculation and now the results are more accurate for particles with a high value for circularity.
RaptIR microscopes no longer incorrectly report that the detector is not optimized
For RaptIR microscopes that did not have a swappable detector, the software sometimes incorrectly reported that the detector needed to be optimized. This has been fixed and you will no longer see this message on these systems.
You can set different scan numbers for each point in the region queue
In earlier versions of software, all points were measured using the same number of scans. Now you can set a separate scan number for each point.
Add comments and custom fields to time series measurements.
Now you can add comments and custom fields to time series data, just like you could with standard measurements previously. To add a comment or a custom field, select Time Series in the measurements pane of the dashboard and then select More to view details. Previously, these fields were not working correctly.
Corrected the area % calculation in MCR analysis
The area percent value was incorrect when doing MCR analysis. This is fixed and now shows the correct area percent.
The stage moves automatically when prompting you to insert or remove the ATR attachment.
When the software prompts you to insert or remove the ATR attachment, the stage now moves automatically to provide clearance.
Fixed the option to use the analyzer when using a polarizer
In previous versions, when using the polarizer with a RaptIR microscope, the No Analyzer option doing the opposite of what was desired. Now it is fixed and selecting No Analyzer will move the analzer out of the beam path.
Known Issues
Repeated prompt asking if you trust the connection to a Nicolet Summit or Summit PRO spectrometer
When connecting to a Nicolet Summit or Summit PRO spectrometer with a built-in computer, a prompt may appear asking if you trust the connection. This prompt should appear only once and then remember your response. However, there is currently an issue that may cause this prompt to appear every time you start the software and connect to the instrument.
Thermo Software IQ reports missing files after upgrading from version 2.3
If you are upgrading from version 2.3, Thermo Software IQ may show that there are missing files after installing the new version. To fix this issue, repair the installation using the operating system tools. When the OMNIC Paradigm software installation screen opens, select Repair.
You may encounter an error during installation.
If you see a message during the software installation that says an error has occurred during installation of OMNIC Paradigm, click Yes to cancel the remainder of the installation. Restart your computer and then start the installation again. This should resolve the issue. If the issue persists, contact a support representative.
Canceling the low-magnification mosaic stops all automation
If you cancel the low-magnification (such as 4x) mosaic, the system should still switch to the high-magnification (such as 15x) objective and perform autofocus. Instead, it does not switch or autofocus automatically. To bring the sample into focus, change objectives in the software and use the autofocus tool.
OMNIC Paradigm v2.4
Release Date: July 2023
Supported Operating Systems: Windows 11 64-bit; Windows 10 64-bit
New features in OMNIC Paradigm software
General Features
Support for FT-Raman with the Raman module
OMNIC Paradigm software now supports FT-Raman measurements using the Raman module with a compatible spectrometer.
Bundle log files
Use the Gather Log Files tool to quickly package your log files and share them with a support representative. This will help us better diagnose and resolve any issues.
Clear all annotations
You can now delete all spectral annotations with just one click, instead of having to manually remove each annotation one by one.
Reset spectrometer
Use can use the Reset Spectrometer tool to move internal components back to the default position and more easily return to an ideal state.
Time Series Measurements
Identify peaks in Time Series data
Like Mercury GC in OMNIC software, the Identify Time Series Peaks search in OMNIC Paradigm software helps you identify materials at different points in your time series measurements.
New report template for Identify Time Series Peaks search
With the new report template, you can save and print your Identify Time Series Peaks search results.
Post-processing atmospheric suppression for Time Series measurements
You can now apply atmospheric suppression to time series data after the measurement. Previously, you had to apply the correction during the measurement.
Workflows and Packages
Sub Workflow tile
With the Sub Workflow tile, you can run another workflow as part of your main workflow, and you can send and receive specific data between them.
Use Workflow variables in the name of template reports
Use a variable in the name of a template replate in your workflows to keep your reports organized and easy to find.
Microscopy
The live spectrum view displays the signal min/max
Now you can see the signal maximum and minimum values in the live spectra view.
The software can detect if the microscope is no longer connected (unplugged or turned off)
Previously, the software might not recognize if you suddenly lost connection to the microscope.
Can close the session from the close session dialog
We added a close button to the Close Session dialog to make it easier to end your session.
Resolved Issues
Time Series Measurements
Now you can import TGA files from Mettler Toledo instruments
Before, you could only import TGA files from TA Instruments products.
Microscopy
The mosaic opacity image slider now works correctly with 4x and 15x mosaic images.
The slider was not correctly blending these different mosaic images, but now it works correctly.
You no longer see errors related to the RaptIR microscope when it was not selected as the active sampling location.
Before, the software would sometimes show error messages related to the microscope even when the microscope was not in use as your active sampling location.
Corrected an issue causing missed scans
During the measurement, stage movement would occasionally cause missed scans in line and area regions. We have introduced some changes to prevent these missed scans.
Known issues
General Features
You must prepare your database before restoring a backup created in v2.2 or earlier
If you are using the default database and have a backup created in OMNIC Paradigm v2.2 or earlier, you must prepare your database before you can restore the backup. See Preparing to restore database backups from earlier versions of OMNIC Paradigm at https://knowledge1.thermofisher.com for instructions.
Selecting Single Beam as your final format can prevent some navigation
If you have your Final Format set to Single Beam, you may see an error indicating that navigation is blocked when you try to view Search Setup or QCheck setup. In the Touchscreen view, you may be blocked from navigating to some other tabs. This is expected because this format is not compatible with those features, but the message and navigation errors are incorrect. Change your final format to another option to use these features.
Microscopy
After canceling a mosaic on the 96 well plate it never switches to the 15x to autofocus
When you cancel the mosaic capture, the system should switch automatically to the 15x objective and begin autofocus. When you are capturing a 96-well plate mosaic, the system does not do this and you must switch manually in the software and then bring your sample into focus.
OMNIC Paradigm v2.3
Release Date: April 2023
Supported Operating Systems: Windows 11 64-bit; Windows 10 64-bit
Before upgrading to version 2.3
Back up your database before upgrading
If you are using the default MariaDB database, back up your database using the Database Backup tool in the software before upgrading. Version 2.3 of OMNIC Paradigm software upgrades the version of MariaDB, so you should have a backup available in case you have any problems with this upgrade and need to recover your data.
If the installation fails during the Instrument Support step, select Yes to cancel the remainder of the installation. Return to the beginning and attempt to install the software again. This should resolve the issue. If it does not, contact technical support or your sales representative.
New features in OMNIC Paradigm software
Import qualification standards
With the new qualification standards tool, you can import DST and QDF standards files and track expiration dates, serial numbers, and more conveniently in the software.
Straight line
Use the straight line tool in the Processing menu to replace a region of the spectrum with a straight line. As with Blank Region, this is useful for removing unwanted features of your spectrum.
Post-processing atmospheric correction
With single measurements, you can now apply atmospheric correction after the measurement is complete. Before, you could only apply the correction before measuring data.
Add many spectra to a library at once
You can now add many spectra to a custom library at once. Before, spectra could only be added one at a time.
NIR Sample compartment qualification workflows
The software now features PHEUR and PV qualification workflows for the NIR Module.
Resolved Issues
Database backups were taking up extra disk space
In previous versions of the software, backing up the database would create extra, duplicate files rather than overwriting the existing backup files. These duplicates could be very large files that would eventually fill your disk space. This is now fixed so that when you back up your database, the new data replaces the old data rather than creating extra files.
Reports now show signatures when you are using Security Suite software
Reports now show digital signatures used with Security Suite software.
The lightbar on Summit spectrometers connected with by USB was not responsive
The Lightbar was not responsive to commands from the software if the instrument was connected with USB. The Lightbar now works correctly over USB.
Changes made to library reader to account for older libraries with different data structures
Some older libraries used in OMNIC had a different data structure that was not supported in OMNIC Paradigm software. We have corrected this issue so that these libraries can now be used.
Added username to reports
The windows account username is included in the reports generated in the software.
We fixed a bug in Advanced Spectral Math
A bug in Advanced Spectral Math prevented you from calculating A/B. This worked in regular spectral math but not Advanced Spectral Math. This has been fixed.
New features and resolved issues for FTIR Microscopy
New features
Measure Time Series data
Now you can measure Time Series data. With a Time Series analysis, you can measure a series of data to identify and analyze change over time, such as in a TGA or GC analysis.
Identify components in time series data
Use Search in your Time Series analysis to identify components in your data. This search is similar to Mercury TGA in OMNIC software and uses MCR and multi-component search to show you what components appeared during your analysis and when they were present.
Import TGA data
If you are measuring TGA data on a TA Instruments analyzer, you can import a TGA file to add weight, temperature, and derivative weight to your profiles.
Single point measure on demand
Added the ability to measure a spectrum at any time and add it to the dashboard. A button was added to the Live Spectrum view that will start a single point collect. The resulting spectrum can be found on the dashboard in the Measurements section.
Backgrounds are now placed at the reticle when the background tool is selected
When the background tool is selected, a background point is automatically placed in the center of the field of view. This allows the user to quickly select a background they have found before they touch the background tool.
Added "return to mosaic" option when using the slide holder reference background location
When using the fixed background reference location, an option has been added to the right hand menu that returns the stage to the center of the initial mosaic.
Support for other objectives
Added support for 10x and 40x optical objectives as well as a Grazing Angle objective, and Diamond ATR.
Resolved issues
Enable "measure background once" for points.
With measure background once selected on the dashboard, after an initial measurement, any subsequent point measurements no longer trigger a new background to be measured.
Improvements to stage control
Improvements have been made to address occasional stage timeouts.
No duplicate request to sign when renaming session
When renaming a Session file with Security Suite software, a duplicate request to sign the file has been removed.
RaptIR error states no longer reported until RaptIR beampath is selected
Camera, stage, and other RaptIR specific error or status dialogs are no longer checked until the RaptIR beampath is selected.
Imported sessions displayed correctly
Fixed situation where imported sessions may not be displayed. Note: large session files still take time to import and will not appear until that import has completed.
Known Issues
Cannot apply atmospheric suppression on Time Series and microscopy session measurements after data measurement
Currently, you can apply atmospheric suppression from the Processing menu to single measurements after they are measured. However, you cannot use this feature with measurements that you collected as part of a Time Series measurement or from a microscopy session. These measurements record a different set of data and do not have the information required for atmospheric suppression. Note that you can use atmospheric suppression during these measurements. You just cannot apply it after the measurement is complete.
OMNIC Paradigm v2.2
Release Date: September 2022
Supported Operating Systems: Windows 10 64-bit
New features in OMNIC Paradigm software
Time-Series Analysis
In version 2.2, OMNIC Paradigm software introduces Time-Series analysis. Time-Series analysis is useful for exploring how sample measurements change over time. For example, you can open a collection of standard measurements to see if your measurements remain consistent throughout a given week or month, helping you identify trends in your data. In version 2.2, you can analyze data in Time Series but you cannot yet collect data directly as a Time-Series Analysis.
Offset scale
Use Offset Scale in the Spectra view to offset your spectra somewhat in an overlay view. Offset scale can help you visually compare peaks.
New features for FTIR microscopy
The Live Spectrum control has been moved from the camera view to the toolbar.
The Live Spectrum control is now in the toolbar and easier to access. When youre using an IR capable objective, use the Live Spectrum control to toggle the live view of the spectrum.
Resolved Issues
Historical results table in workflows
The historical results table in workflows was not correctly showing all results. This is now fixed.
Session files are now backed up and restored when you back up or restore the database
Known Issues
Display scale resets to Full XY Scale after exiting zoom
Previously, only the portion of the session data that was saved in the database was backed up. Now, the entire session is backed up and restored when you back up and restore your database. In the spectra view, the display resets to Full XY Scale after zooming in on a portion of the spectrum.
OMNIC Paradigm v2.1
Release Date: July 2022
Supported Operating Systems: Windows 10 64-bit
New features in OMNIC Paradigm software
Contaminant analysis: Identify contaminants in your sample
Use Contaminant Analysis to quickly identify and quantify potential contaminants in your sample. You provide a known material, and OMNIC Paradigm tells you what potential contaminants are in your sample.
Data smoothing: Reduce noise in your spectra with data smoothing
Use Smooth in the Processing menu to reduce noise in your spectra. Apply either automatic smoothing for fast improvements or fine-tune the smoothing by adjusting how the Savitsky-Golay algorithm is applied.
Normalize: Make comparison simple by normalizing the scale of spectra
You can now change the Y-axis scale of spectra to a "normal" scale, in which the Y values of the data points range from 0 absorbance units for the lowest point to 1 absorbance unit for the hightest peak or from 10% to 100% transmittance. These normal scales are typical of spectra in commercial libraries.
Clean Up Database: Keep your records tidy with the database cleanup tool
With the new database cleanup tool, you can automatically delete old measurements when they reach a preset age.
Search setup shows the current spectrum
If you open the Search Setup from the Spectral view, the region selection now shows your currently selected spectrum instead of the default example spectrum, making it easier to specify regions around peaks in your specific spectrum.
Filter records by signature status (with Security Suite software)
If you are using Security Suite software, you can now use the Signature filter to find records based on their signature status. For example, you can now quickly find all your records that need to be approved and approve them all at once.
Print the system status
Now you can print your system status from the System Status dialog.
New features for FTIR microscopy
Polarizers are enabled
If your Nicolet RaptIR FTIR microscope includes the optional polarizers, you can now control the visible and IR polarizers in the software.
QNT methods can be used as a profile type
You can now use a quant method created in TQ Analyst software as your chemical image profile in FTIR microscopy. The quant profile can be applied only after collecting data.
Automatically move the stage to a background reference position
When using the RaptIR sample holder, you can automatically move the stage to the built-in reference location for easier background collection.
Switch between three objectives
If you have three objectives installed, you can now easily cycle between them or choose a specific objective.
Additive mosaics
Move the stage and collect a new mosaic (high or low magnification) and add it to your mosaic sky view. Makes it easier to example a large area of your sample or to focus on a new portion of the sample.
Analyze MAP files from OMNIC software
Open .map files created in OMNIC software and automatically convert them to a .session file that you can explore in OMNIC Paradigm software.
Import / Export RaptIR Settings
Now you can export and import your saved settings used with the Nicolet RaptIR FTIR microscope, just as you could for spectroscopy settings.
Delete sessions to clear space on your system
Previously, you could not delete a session. Now you can right-click a session on the dashboard and delete it.
View the exact stage position
In the microscopy session view, you can now see the exact X, Y, and Z stage position.
Resolved Issues
Workflow editor: Sample prompt screen was not able to display a PDF
Previously, if you added a PDF to the Sample Prompt screen in a Measure workflow tile, the PDF would not be saved with the prompt and could not be displayed. Now the PDF is correctly attached to the Sample Prompt.
Workflows: The Equation tile and Variable tile would break if you copied or renamed a workflow
Variables that used the workflow name were not updated if you copied or renamed the workflow, which caused the tiles to break. Now the variables are automatically renamed to match the updated workflow name.
If you created workflows in earlier versions of the software, you need to update your Equation and Variables tiles manually before the variable can be renamed automatically. To update the tiles, open them and replace any variables so that they use the new workflow name. Then close the tile and save the workflow.
When you have an accessory installed, the Settings list on the dashboard shows only the settings that were created with the matching accessory.
Previously, the list showed all settings, including those that were created for a different accessory and not useable by the accessory you have installed. Now the system only lists settings that were created for the currently installed accessory.
We repaired Restore Database, so you can once again restore from a previously backed up database.
With the 2.0 version of OMNIC Paradigm software, you could back up your database but not restore it. Now, Restore Database is once again working correctly.
Known Issues
The database backup tool backs up only a portion of session data.
Session data contains two parts: a portion of the session is stored in the database and a portion of the session is saved in files on the disk drive. The database back up tool in OMNIC Paradigm software backs up only the data stored in the database. To ensure you can restore your data, you must use a backup utility program to back up the files stored in the following directory:
C:\Users\Public\Documents\thermo scientific\omnic paradigm\Sessions
The polarizer cannot be used to measure lines, only areas and points.
Currently, the polarizer can be used to measure areas and points, but not lines. If you draw a line and then enable the polarizer, the line will be measured with the polarizer at only the fixed, starting position.
ATR settings must be adjusted and saved for each new session
Currently, every time you start a new microscopy session using ATR analysis, you must open the ATR Contact dialog, change the settings, and then save the changes.
RaptIR qualification workflows may fail if the polarizers are in the beam path
If your microscope has the optional polarizers, make sure they are moved out of the beam path before you run the qualification workflows. If they are left in the beam path, the data will be poor and the tests may fail.
A new background measurement is collected before measuring points
When measuring points, a new background measurement is always collected before measuring any points, even if you accepted and collected a background measurement manually. The correct location is used, but the system performs an extra step.
Returning to the dashboard returns the visual polarizer to the default settings
If you return the dashboard during your microscopy session, the visual polarizer settings are returned to their default position.
OMNIC Paradigm v2.0
Release Date: February 2022
Supported Operating Systems: Windows 10 64-bit
New Features
New FTIR microscopy tools and support for Nicolet RaptIR FTIR Microscope
OMNIC Paradigm software now includes a new interface and set of tools for FTIR microscopy with the Nicolet RaptIR FTIR Microscope.
Support for Nicolet iS50 spectrometer and common sampling accessories
You can now use OMNIC Paradigm software to configure and run the Nicolet iS50 spectrometer and sampling accessories. Supported accessories include the built-in ATR module, the Smart iTR, the Smart Diffuse Reflectance, the Smart NIR Integrating Sphere, the Smart Golden Gate, the Smart iTX, the Smart ARK, Smart OMNI-Sampler, Smart SAGA, and the Smart OMNI-Transmission.
Better multi-tasking with tabs
The spectra and session views support a tabbed view, so you can open measurements in separate tabs.
Workflow enhancements
The Measure tile in Workflows now includes more instrument and measurement settings, including gain, aperture, sample spacing, and zero fill.
Resolved Issues
Operator view: Workflows report that a workflow is already running
When using the Operator view, if you tried to start a workflow, the system would sometimes show that a workflow was already running in the background.
Optical Resolution tests failing in CP, JP, and Factory Qualification workflows
"A change was made to the Optical Resolution tests in the Nicolet FTIR - CP Qualification, Nicolet FTIR - JP Qualification and Nicolet FTIR - Factory Qualification workflows. This change makes the measurement immune to baseline shifts caused by variations in the finish of the Polystyrene standard. This improvement to the measurement is consistent with the approach used in OMNIC ValPro tests. Customers upgrading from OMNIC Paradigm v1.9 will observe larger reported values for these tests after upgrading to OMNIC Paradigm v2.0. This is expected. All previous results are valid. This change is just an improvement to the measurement."
Known Issues
Measurements collected in a workflow may show an invalid signature
Occasionally, when measuring and signing data in a workflow, the signature event may incorrectly occur and be saved before the measurement event. This can happen because all measurement data is stored at once, rather than in a specific, predefined order. If this happens and the data is saved out of order, the measurement shows an invalid signature since it appears that the data was changed after it was signed.
When collecting very large maps, the computer may enter power saving mode and suspend the USB devices
If you are collecting large maps, such as large area maps, the computer may enter a power saving mode and suspend the USB devices, interrupting the collection and disconnecting the mouse and keyboard. To ensure that the collection finishes, disable USB selective suspend settings in the Windows power options.
Cannot access QCheck Setup in the Touchscreen interface
When viewing a spectrum in the Touchscreen interface, the menu item for QCheck Setup appears enabled but does not open the setup screen. To edit your QCheck settings, select QCheck from the Home screen.
OMNIC Paradigm 1.9 HF2
Release Date: October 2021
Supported Operating Systems: Windows 10 64-bit
Resolved Issues
Improved services for distributed installations of OMNIC Paradigm software with Security Suite software
Modified service startup logic for the IR Instrument Engine to ensure that the service responds to the Windows service control manager in a timely manner.
OMNIC Paradigm v1.9 HF1
Release Date: October 2021
Supported Operating Systems: Windows 10 64-bit
Resolved Issues
Workflows were returning incorrect results on systems using certain language settings
In version 1.9, if you were running your system using languages that use the comma as a decimal separator, workflows would return incorrect results.
OMNIC Paradigm v1.9
Release Date: October 2021
Supported Operating Systems: Windows 10 64-bit
New Features
Use Reprocess in the Process menu to transform the original interferogram data for your spectrum with new settings
With Reprocess, you can transform your original data to better match library or other reference spectra, giving you a more accurate comparison.
With Blank Regions in the Process menu, you can clear unwanted data from your spectrum
Blanking a region is useful when your spectrum contains unwanted noise that would otherwise interfere with your analysis. Use Blank Regions to clear, or delete, a region of the spectrum. Blank Regions is also available in the Processing tile in Workflows.
You can now calculate average and standard deviations of spectra using the Statistical Spectra tool in the Process menu
With Statistical Spectra, you can generate new spectra that represent the average or standard deviation of 2 or more spectra. You can also use Statistical Spectra in the Processing tile in Workflows.
Add custom logos to reports and template reports
Go to Configure > Options to use your own company logo in reports and template reports. Your logo replaces the default Thermo Scientific logo on any custom reports you create and any template reports created in a workflow.
Workflow enhancements
Find Peaks, Blank Regions, and Statistical Spectra are now all available in workflows.
Enhanced system status information
The system status information has been updated to provide more accurate information. New icons indicate if your system is using security or not, clearer messages describe connection status, and updated messages help you troubleshoot any potential issues.
Thermo Scientific Web API Server requires inbound connections on TCP Port 9004
The new Thermo Scientific Web API Server is required for instrument connections. Make sure your firewall settings allow inbound connections over TCP Port 9004 on Private, Public, and Domain profiles.
Resolved Issues
Spectra in workflows are automatically deresolved to match quantification methods
Previously, in a workflow, if the resolution of your spectrum did not match the data in a quantification method, the method would fail. Now, the resolution of the spectrum is automatically lowered, or deresolved, to match the quantification method.
Known Issues
You cannot reprocess older sample measurements that were collected in a workflow
In OMNIC Paradigm software versions 1.8 and earlier, any sample measurements collected in a workflow stored interferograms incorrectly. Reprocessing these measurements will result in incorrect results. Measurements that you collect in version 1.9 and later can be reprocessed correctly.
When installing OMNIC Paradigm with Security Service pack, the installer incorrectly attempts to install Audit Manager software on the client
To install Paradigm with Security Service Pack on a distributed installation, run Start.exe on DVD 1 on the server. When prompted, select initial installation. On the client, go to Apps and Features and uninstall Audit Log Service, if it is present. Then, run Start.exe on DVD 1. When prompted, select Subsequent Instruments. When DVD 1 is complete, run Start.exe on DVD 2.
OMNIC Paradigm v1.8
Release Date: June 2021
Supported Operating Systems: Windows 10 64-bit
New Features
Support for more Quantification Methods
Now you can use five more quantification methods in OMNIC Paradigm software, including
-
QC Compare
-
Search Standard
-
Distance Match
-
SMLR (Stepwise Linear Regression)
-
PCR (Principal Component Regression)
See the help information TQ Analyst software for more information on these quantification methods
Copy and paste spectra to other applications
Copy and paste spectra from the spectra view to external applications, including office applications or image editing software.
More security controls for creating and managing settings
If you are using OMNIC Paradigm software with Security Suite applications, you can now set Access Controls for the New, Rename, Save, and Delete buttons for measurement settings. There are now also audit events for logged for these and other modifications. You can also control permissions for the More button to restrict access to advanced settings.
Resolved Issues
Multi-component search was not respecting regions
If you set limited regions in your Search Setup for multi-component search, the entire range was searched. Now, multi-component search correctly respects the specified range.
Multi-component search report now shows the selected composite result
Previously, reports for multi-component search results showed only the first composite result. Now, the report shows whichever composite result you have selected when you create the report.
Known Issues
OMNIC Paradigm software no longer supports Oracle 12c Release 2 or 18c for the database engine.
If you are already using Oracle 12c Release 2 or Oracle 18c for your database engine, you can continue to use them, but you should consider upgrading to Oracle 19c for the best support. See Oracles help documentation for more information on upgrading your database.
If you have upgraded from Paradigm 1.6 (or earlier) to 1.7 (or newer), you may be unable to link your instrument to Thermo Fisher Connect.
If you have an issue linking an instrument after an upgrade:
-
Delete these folders if they exist:
C:\ProgramData\Thermo Scientific\Paradigm\deeplaser\
C:\ProgramData\Thermo Scientific\Paradigm\deeplaser-1.8.4\
-
Reboot the computer.
OMNIC Paradigm v1.7
Release Date: April 2021
Supported Operating Systems: Windows 10 64-bit
New Features
Support for the Nicolet Summit OA spectrometer
OMNIC Paradigm software now supports the Nicolet Summit OA FTIR spectrometer. The software comes pre-installed on all Summit OA spectrometers.
Link your instrument to your Thermo Fisher Connect account
Use the Instrument Health feature in the software to link your instrument to your Thermo Fisher Connect account.
Run an Executable file from within a workflow
With the Run Program tile, you can run an EXE file from within a workflow. Pass arguments to the EXE file and use any returned values as variables in your other workflow tiles.
Resolved Issues
The software notifies you if the security services connection is lost
It was possible for the software to become disconnected from security services running in the background without providing any notification. Now, if the connection is lost, the user is notified and is forced to exit the software.
Audit Manager software reported that a workflow was Finished even if it was exited early
When canceling a workflow early, the Audit Manager software displayed Finished in the audit event. Instead, it now shows Cancelled if the workflow was exited early.
Custom reports created in workflows now correctly encode all characters
Custom reports created with a workflow did not correctly encode some characters. These reports now correctly use UTF-8 character encoding. The fix does not correct previously created reports, but it does apply to your current workflows.
Known Issues
You cannot use region selections with a multi-component search
Multi-component search does not use any regions you specified in search setup. The regions correctly apply to a correlation search, but not to a multi-component search.
OMNIC Paradigm v1.6 HF1
Release Date: February 2021
Supported Operating Systems: Windows 10 64-bit
Resolved Issues
Cloud Connector could not fully connect
Several files needed by Cloud Connector were not installed. We have restored these files so that Cloud Connector can fully connect.
OMNIC Paradigm v1.6
Release Date: December 2020
Supported Operating Systems: Windows 10 64-bit
New Features
Custom labels and annotations
You can now add custom annotations and peak annotations to spectra. Your annotations are displayed on any reports you create.
Add tables to custom reports in workflows
Add a table of results to custom reports in your workflows. The table displays results from the current run and from previous runs.
Derivative in the Processing tile
Calculate first and second order derivatives with the Processing tile in workflows.
Run a workflow automatically
Use the Auto Run feature to schedule a package workflow to run automatically in Operator mode.
Resolved Issues
Axis labels were too big on exported reports
Labels on reports were sometimes too large and overlapped the data. The labels are now a more appropriate size.
The handles to adjust your selection with Find Peak were too small
We increased the size of the handles to adjust the Find Peak region so that they are easier to use.
Default file path changed from Schema to Paradigm
Previously, packages and workflows used a default file path that included the word Schema. We have updated this to say Paradigm instead. The update does not affect any current data and the system will continue to use your current file location.
There was not a notification when the database became disconnected
If the database became disconnected while you were collecting data, there was no notification letting you know that your data was not being saved. Now, a notification alerts you that you need to restore the database connection. In Operator mode, you are prevented from running a workflow while the database is disconnected.
Known Issues
You cannot define custom colors to use with annotation tools
You can choose colors to use with annotation tools, but you cannot currently specify your own color. The Define Custom Color tools are disabled.
Align and Calibrate tools are incorrectly disabled
The Align and Calibrate menu options are sometimes incorrectly disabled after previewing a background or viewing a spectrum. If the options are disabled while you are viewing the dashboard, click Dashboard in the toolbar again (even if you are already viewing the dashboard) to enable Align and Calibrate.
OMNIC Paradigm v1.5 HF1
Release Date: November 2020
Supported Operating Systems: Windows 10 64-bit
Resolved Issues
The software did not always close after running workflows
We fixed an issue that prevented you from exiting the software after running a workflow.
Updating the software caused multiple versions to appear in the list of installed applications
After updating the software, you may have seen multiple versions in your list of installed applications. Now only the latest version appears.
OMNIC Paradigm v1.5
Release Date: September 2020
Supported Operating Systems: Windows 10 64-bit
New Features
Support for the Nicolet Summit LITE spectrometer
You can now connect to and operate a Nicolet Summit LITE spectrometer.
Develop and Run Custom Solutions with the Package Editor and Operator Interface
Use the new Package Editor to bundle workflows and their required files into RDEP files. Open and run package workflows in the Operator interface.
Spectral Math in Workflows
The Processing tile in Workflows now supports spectral math, including addition, subtraction, multiplication, and division using two spectra.
Measurements and Reports Created in Workflows Support Digital Signatures
When using Security Suite software, measurements and reports created in workflows now support digital signatures.
Resolved Issues
When Using a Discriminant Analysis Method with the TQ Analyst Tile, Class Names are Now Available as Variables.
Previously, Class Names from discriminant analysis were not available as variables which meant that some results could not be used in other tiles, such as the Decision tile.
The Match Settings Option in the Measure Tile Now Correctly Matches Zero Fill
When using the Match Settings option in the Measure tile, the settings did not correctly match the Zero Fill setting. We fixed this so the Zero Fill is correctly matched.
You can now Install Paradigm successfully on Non-English versions of Windows.
If you installed OMNIC Paradigm software on a non-English version of Windows 10, the software would hang when you tried to measure a sample. This is resolved and you can now run the software as expected on non-English versions of Windows 10.
Spectrum History Information Displays Correctly after Importing a SPA File that Was Created on a System using Double-Byte Characters.
The spectrum history information did not display correctly when importing a SPA file from a system that was using double-byte characters. The history is now correct if your system is also using double-byte characters. The history information may still have display problems if your system is using single-byte characters and you import a SPA file from a system using double-byte characters.
Known Issues
The Library Title Does Not Display double-byte Characters Correctly in the Library Manager.
When running the software with a system using double-byte characters, the Library Title field in the Library Manager does not display correctly.
Reports May Be Displayed Incorrectly in the Reports Pane
Occasionally, the reports pane will display a duplicate report after creating a new report. Additionally, the report preview image may not be updated and will show a different report than the currently selected report. These are display issues only and do not affect your data. Click Refresh in the reports pane to refresh the list. If this does not solve the problem, restart the computer to clear the duplicate reports.
OMNIC Paradigm v1.4
Release Date: July 2020
Supported Operating Systems: Windows 10 64-bit
New Features
Advanced Spectral Math
While standard spectral math supports spectral subtraction, addition, multiplication, and division using a single reference spectrum, you can now use Advanced Spectral Math to build custom equation using up to 10 reference spectra and using a wider range of powerful operations.
Revert Measurement
You can now revert your measurement to an earlier step in your processing and analysis, allowing you to undo any changes that degrade your data. Previously, you may have had to measure the sample again to have clean, unprocessed data.
Duplicate Measurement
With the new Duplicate Measurement feature, you can make a copy of a measurement from its current state or from an earlier step.
View Measurement History
You can now view the measurement history to quickly visually compare your measurement data at different stages of processing and analysis. For example, the measurement history view is great for quickly comparing the spectrum before and after applying a baseline correction.
Measurement History Report
The measurement history report displays the spectrum at each stage in you processing and analysis so that you have a convenient record of how the data was changed.
Export Spectra from Sub Panes
Previously, you could export only top-level measurements from the Results Pane of the spectral view. Now you can also export sub items, such as reference spectra or spectra used in spectral math operations.
Enhanced Security Features when Paired with Security Suite Software
We added new policies, access controls, and default settings to Security Administration software for managing OMNIC Paradigm. You can now prevent the measurement preview from displaying during measurement or disable the pause button during a measurement and an audit event is logged when a workflow is cancelled.
New Software OQ Procedure for OMNIC Paradigm v 1.4 and later
There is a new, updated Operational Qualification procedure for use with Security Suite software and OMNIC Paradigm software. The updated procedure includes changes to access controls and updated system policies.
Resolved Issues
Custom reports created in a workflow are now displayed correctly in print preview
When printing a custom report created in a workflow, the print preview screen showed a mostly empty page. This is fixed and the print preview now shows the custom report correctly.
The preview image for embedded workflows is now updated correctly
If you used the Embedded Workflow tile, the preview image wouldnt update to show any recent changes to the embedded workflow after you used the Update Embedded Workflow(s) command. The command now correctly updates the preview image so you can see the latest changes to the workflow.
New tags are now displayed immediately when added to a measurement
If you add a tag to a measurement and then immediately click Manage Tags, the new tag now correctly shows up immediately.
You can now import multiple SPA files at once.
Previously, an error occurred if you tried to import multiple SPA files as once. We fixed this so now you can import multiple spectra successfully.
Known Issues
It is possible to exit a workflow when working in a secure environment
If you are running a workflow in the touchscreen interface, the Home button in the menu is still enabled, allowing you to exit the workflow even if the prevent cancel policy is enabled.
OMNIC Paradigm v1.3 HF1
Release Date: May 2020
Supported Operating Systems: Windows 10 64-bit
Resolved Issues
The Spectrum Information Now Shows the Correct Detector Name
The spectrum information showed an incorrect detector name. It now correctly displays DTGS KBr as the detector in the in the Spectrometer tab of the Spectrum Information.
Cannot Click the More button in the Measurement Pane during Laser Calibration
Clicking the More button caused laser calibration to lock up. The button is now correctly disabled during laser calibration.
Commercial Library Spectra Cannot be Exported
When multiple spectra are selected in the Spectra view, only the non-commercial library spectra can be exported to a folder.
The iS5 System Recognizes when an Accessory is Removed
The iS5 system did not always recognize if an accessory was removed, which prevented the Laser Calibration from being performed.
Known Issues
Software Installation Status Icon Shows Simulation
After you install the software, it may show Simulation (Nicolet iS5) in the status icon. If this is the case, use the Connectivity menus Connection command to reselect the iS5. When the switch is complete, ignore the status icon and leave the instrument connected to the PCs USB port. Close the software and reboot the PC. The status icon should now show: Nicolet iS5.
OMNIC Paradigm v1.3
Release Date: March 2020
Supported Operating Systems: Windows 10 64-bit
New Features
Nicolet iS5 Support
OMNIC Paradigm software is now available to be installed on a computer controlling a Nicolet iS5 spectrometer. This software can be installed on any Windows 10 computer. The OMNIC Paradigm iS5 software is also compatible with our Security Suite software and can be used with a Distributed installation.
Discriminate Analysis
Discriminate Analysis quantitative method created in the TQ Analyst can be used in OMNIC Paradigm.
Options
You can now customize the color and line weight of spectra in the spectral display and set the number of digits shown on the display and in peak tables.
Faster Multi-component Search
Multi-component search is now at least four times faster. The total time for the search depends on the number of libraries used in the search, the number of spectra in the libraries, and the processing power of the computer.
Powerful New Workflow Tiles
We added four new, advanced workflow tiles to the Workflow Editor:
-
Variable tile: Create custom variables or modify the value of an existing variable
-
Equation tile: Use built-in functions and custom equations for calculations
-
Embedded Workflow tile: Embed one workflow within another
-
Workflow Table tile: Import a table of values to use throughout the workflow
Report Configuration
You can now configure reports to optionally show spectra, table data, and meta-data.
Translations
OMNIC Paradigm software is now available in Italian. The Japanese translation has been updated.
Security Suite New Features
All new options listed above are included in the Security Suite software updates for the OMNIC Paradigm software. Access Control policies are provided to manage users within the Thermo Scientific Security Administration application.
Resolved Issues
Spectral Name Changes Now Appear in the Spectral History
OMNIC Paradigm software now displays an entry in the spectral history for a spectral name change.
Fixed Crash Caused by Clicking in the Spectrometer tab
We fixed an issue that caused the software to crash if you clicked in the Spectrometer tab of the Spectrum Information.
Correct Number of Scans in Workflows for the French Translation
In the French translation, measurements in workflows allowed only scans of 1, 2, 3, or 4 and the scans entered were multiples by 256. Workflows now allow the correct number of scans.
Zooming in Multi-component Search Upper Pane is Now Reflected in the Lower Panes
The limits on the panes were not in sync when zooming (boxing).
Library Search in Workflows Now Correctly Handles Libraries of Different Ranges, such as Raman and NIR
If you used libraries with different spectral ranges in a search, such as FTIR, NIR, and RAMAN, the result would be No search results found. The search now returns correct results.
In Spectral Math, We Changed the Sample and the Reference Positions
The Result is in the top pane, the Sample is the middle pane, and the reference is the bottom pane.
The Search Tile in Workflows Now Retrieves Libraries from all the Paths Defined in the OMNIC Paradigm Software
Previously, the search tile retrieved libraries only from the default OMNIC Paradigm library path.
Known Issues
Dashboard toolbar disappears and doesnt come back from a Workflow
This issue is caused due to an invalid workflow. Restart the software.
OMNIC Paradigm v1.2
Release Date: November 2019
Supported Operating Systems: Windows 10 64-bit
New Features
Workstation Support
OMNIC Paradigm software is now available to be used on a separate computer as workstation. This software can be installed on any Windows 10 computer. The Workstation software is also compatible with our Security Suite software and can be used with a Distributed installation.
Remote Connectivity
You can now control Nicolet Summit spectrometers remotely. Install OMNIC Paradigm Workstation software on a connected computer and use it to control the Nicolet Summit spectrometer. Connect over WiFi or Ethernet cable. (See OMNIC Paradigm documentation for additional information.)
Database Maintenance: Back Up and Restore
Back up and restore your default Thermo Fisher Scientific Built-in MariaDB by using the new database management tools. Use the Back Up tool to back up the database manually or automatically to a specified schedule. Use Restore to restore a previously saved database backup to the current database location.
QCheck now Supported with Workflows
Run a QCheck analysis with OMNIC Paradigm workflows. In the Workflow Editor, the QCheck tile can be configured with the same settings available in the OMNIC Paradigm desktop software. You can also include QCheck in a workflow when creating a workflow automatically from the Spectrum History Information.
Database Spectra can be Used with Workflows
When building workflows, you can now use the Load Spectrum tile to load data from the database rather than from only a saved SPA data file.
Kramers-Kronig Correction
Kramers-Kronig data correction is now available as a Processing option in the OMNIC Paradigm software and in the OMNIC Paradigm workflows.
Import and Export SPC Format Spectral Data
You can now import and export GRAMS SPC files from the OMNIC Paradigm desktop and touchscreen software. Additional file formats previously supported in OMNIC software can be converted using the TQ Analyst software and then imported into the OMNIC Paradigm software as either SPA for SPC file formats.
New Diagnostic Workflow for Testing ATR Accessories
A new diagnostic workflow is available to test the throughput of your ATR accessories. The workflow will walk you through the test to determine if your ATR accessory is working properly. Contact your Thermo Fisher Scientific Service Engineer for further information.
Translations Available
OMNIC Paradigm software is now available in the following languages: Chinese (Simplified), Japanese, Polish, French, Spanish and Russian.
Side Panel on Desktop Software Now has a Collapse Option
We added an arrow icon to the spectral view in the desktop software that you can use to collapse and expand the side panel.
Display Limits Options
You can now use the right-click menu in the spectral view to modify the display limits by entering the X and Y limit information. This option is available for both desktop and touchscreen.
Spectral Math Can Now be Used by Other Final Formats
Spectral math now works with single beam data.
Titles are Now Available in Overlay Mode of the Spectral View
Spectrum titles are now visible when in Overlay mode in the spectral view.
Security Suite New Features
All new options listed above are included in the Security Suite software updates for the OMNIC Paradigm software. Access Control policies are provided to manage users within the Thermo Scientific Security Administration application.
Resolved Issues
Workflow Generated Spectra with Attached Interferograms
OMNIC Paradigm software now handles interferograms attached to data that was collected with OMNIC Paradigm workflows similarly to data collected with the desktop and touchscreen versions of OMNIC Paradigm.
Clear Sample Name with Backspace Key
When changing the sample name using the backspace key, the previous name was not cleared properly.
Collect Background No Longer Disabled in Touchscreen
The Collect Background option is no longer disabled when the number of background scans is set to fewer than 10.
Create Report Not Working as Expected
We fixed issues that were causing Create Report to display different scaling and display limit information.
Spectrum Collection and Processing History was Not Capturing Title Change
Spectrum Collection and Processing history now shows any title changes for the spectral data record.
Select All was Not Processing All Selected Spectra
Select All was previously not operating on all selected spectra for processing functionality.
Removed Extra Dialogs that Appeared When Inserting and Removing Smart Accessories
We removed unnecessary dialogs that were appearing in the touchscreen interface when smart accessories were inserted or removed.
Collection & Processing Information for Zero Fill Reports Correct Values
Spectra collected using a workflow were reporting Unknown for the Zero Fill value in the Collection & Processing Information, but now display the correct zero fill values.
Number of Background Scans Value Not Set Properly
When trying to set the number of background scans for background options Measure every _ minutes and Measure before sample, the value could not be less than 10 and the tooltip was incorrect. These issues have now been corrected.
Smart Background Scans
Smart Background sometimes collected more scans than specified. It now collects the correct number of scans.
Improved Scroll Bars in Touchscreen Interface
We improved the scroll bars in the touchscreen interface to make them consistent and easier to use.
Peak Labels no Longer Cut Off When Printing
Peak labels displayed on the spectral display are no longer cut off when printing.
Hide All Now Hides All Spectra
The Hide All option on the spectral display side panel did not always hide all spectra. This is now fixed.
Import SPA and SPC Files From a Network Location on an Instrument
Supported files can now be imported from both the local computer and from a specified network location.
Improved Speed to Index Large Amounts of Data
Indexing large amounts of data in the database was very slow but is now much faster.
Fixed Text Search with User Libraries
Text search sometimes failed when searching User Libraries, but it now works consistently.
Known Issues
Spectral Display for Interferogram, Single Beam or Final Format spectrum under More button on Dashboard can appear cut off
This issue can be solved by modifying your Display Settings for your computer. Under Display Settings in the Windows Settings there are two settings that need to be set to the recommended values:
-
Resolution must be set to the recommended value
-
Change the size of text, apps and other items needs to be set to 100%.
This may also require a reboot of the computer to get the display to show properly
OMNIC Paradigm v1.1
Release Date: July 2019
Supported Operating Systems: Windows 10 64-bit
New Features
Security Suite Support
OMNIC Paradigm software is now supported with the Security Suite software in both Single computer installation and the Distributed installation. Administration of policies is controlled using Security Administration and the Audit Trail information can be found in Audit Manager.
Update Feature
You can now monitor a network location or USB location for updates so that you can be notified automatically of new software releases.
Resolved Issues
Localization
We fixed localization problems that were introducing errors throughout the software, including the System Status dialog.
Cloud Connector Enhancements
Improved the Cloud Connector to make it more robust so that it can better handle when the internet connection is inactive or lost.
Gain and the Interferogram
The interferogram is now automatically updated when you change the gain settings.
Usability improvements for Touchscreen
You can now toggle and scroll the main menu when using the touchscreen interface.
Database Configuration (Touchscreen Interface)
If you cancel your changes when configuring your database, the changes are now discarded. Previously, unwanted changes were saved.
Confirmation Dialog Added when You Delete a Workflow
We added a confirmation dialog to verify that you want to delete a selected workflow before it is deleted.
Clear Spectrum from right click menu only cleared last spectrum when all spectra are selected
When all the spectra in the window are selected and Ctrl W or Clear Spectrum is used from the right click menu, only the last spectrum was cleared instead of all the spectra. Now, all the spectra are cleared from the display.
Acquire Menu Items No Longer Disabled
We fixed an error that was causing all items in the Acquire menu to be disabled if you ended a workflow early.
Missing Checkbox in the Touchscreen Interface
A checkbox you would use to specify the number of background scans in not present in the touchscreen interface. The checkbox is present in the desktop interface.
Known Issues
Run Workflow Button is Incorrectly Disabled
If the instrument is not ready, the Run Workflow button may be disabled.
History is Not Showing Correlation Search or Multi-Component Search Information
Spectral histories are missing information for Correlation Search and Multi-Component Search. This information does appear in the Collection and Processing information.
Changing Library Location Requires an Update to Workflow Search Tiles
If you modify the location of a library that you have referenced in a Search tile in a workflow, you must update the Search tile again before running the workflow.
OMNIC Paradigm v1.0
Release Date: May 2019
Supported Operating Systems: Windows 10 64-bit
New Features
Initial release
This is the initial release of the OMNIC Paradigm software.
Resolved Issues
None - Initial version
Known Issues
None
How to Contact Us
Current contact information is located at www.thermofisher.com
Select the Contact Us icon at the top of the screen
Trademarks
Microsoft and Windows are either trademarks or registered trademarks of Microsoft Corporation in the United States and/or other countries.
All other trademarks are the property of Thermo Fisher Scientific Inc. and its subsidiaries.
Attachment(s):
| File | Last Modified |
|---|---|
| OMNIC Paradigm software release notes.pdf | September 10, 2025 |