How to Run OMNIC 9.8 in Simulator Mode
To run Omnic 9.8 in simulator mode, first modify the OmnicSettings.ini file by setting simulator=1 in the [Bench_CFG] section. Then, install the OMNIC Instrument Config application by executing the OMNIC Demo Instrument Config.msi file found in the InstrumentDemo folder. This application allows you to select the Instrument Simulator and switch between simulating the iS50 and the iS5, with options for Basic OMNIC or Advance OMNIC for iS5 simulation.
Description:
The attached procedure describes how to run OMNIC 9.8 in simulator mode.
Environment:
OMNIC 9.8
Attachment(s):
| File | Last Modified |
|---|---|
| How to Run OMNIC 9.8 in Simulator Mode.pdf | December 15, 2021 |