How to Change the Service Account that Controls the OMNIC Secure Data Folder
To change the service account controlling OMNIC’s secure data folder, assign the new account full access to key secure subfolders, update the login credentials for the related services, and restart them to complete the transition.
Description:
The attached document describes how to change the service account controlling the OMNIC secure data folder.
Environment:
OMNIC
Attachment(s):
| File | Last Modified |
|---|---|
| How to Change the Service Account that Controls the OMNIC Secure Data Folder.pdf | December 15, 2021 |