Specta with Security Does Not Show .SPA Files in the File Open Dialog Box
Description:
When OMNIC Specta v2.2 is installed with Security Suite v2.0 or higher, it will not show any .SPA files in the File Open dialog box. When the dialog box first opens, it will only display .SPG files contained within in the folder.
Environment:
Omnic Specta
Attachment(s):
| File | Last Modified |
|---|---|
| Specta with Security Does Not Show .SPA Files in the File Open Dialog Box.pdf | July 29, 2022 |