Stopping Data Collection Prior to Completion of Full Number of Scans
Users can stop OMNIC data collection early by pausing the scan, closing the spectral window, and declining to save the spectrum if prompted. OMNIC will then be ready to begin a new scan.
Description:
Some situations it may be helpful to terminate a data collection prior to the completion of co-adding the the full number of scans. The attached document describes the process for stopping data collection.
Environment:
OMNIC
Attachment(s):
| File | Last Modified |
|---|---|
| Stopping Data Collection Prior to Completion of Full Number of Scans.pdf | December 16, 2021 |