Transferring OMNIC Audit Trail Information from SQLite to SQL Server
To migrate OMNIC audit trail data from SQLite to SQL Server, export audit events to CSV using SQLite tools, then import into SQL Server via the Import Wizard, ensuring proper formatting and data types.
Description:
This document describes the process of manually transferring audit events from SQLite to the new SQL Server database.
Environment:
Audit Manager database with OMNIC Security package
Attachment(s):
| File | Last Modified |
|---|---|
| Transferring OMNIC Audit Trail Information from SQLite to SQL Server.pdf | December 25, 2021 |