ValPro for Dispersive Raman
ValPro for Dispersive Raman ensures the accuracy and reproducibility of DXR spectrometers using a traceable polystyrene standard. Compliance services include installation, certification, and recertification. Diagnosing qualification failures involves adjusting stage focus, aligning the spectrometer, and performing calibrations. The ValPro package supports various lasers and gratings but excludes high-resolution gratings.
Description:
The attached document provides basic information and installation instructions for ValPro for DXR.
Environment:
Raman DXR
Attachment(s):
| File | Last Modified |
|---|---|
| ValPro for Dispersive Raman.pdf | December 17, 2021 |