OMNIC Specta Release Notes
OMNIC Specta Release Notes
About This Document
This document contains a revision history of OMNIC Specta, including new features that may not be included in the User’s Guide, resolved issues, and known issues.
The following conventions are used in this document:
Trademarks
Microsoft, Internet Explorer, and Windows are either trademarks or registered trademarks of Microsoft Corporation in the United States and/or other countries.
ToolBook is either a trademark or registered trademark of SumTotal in the United States and/or other countries.
Semprex is either a trademark or registered trademark of Semprex Corporation in the United States and/or other countries.
PerkinElmer is either a trademark or registered trademark of PerkinElmer, Inc. in the United States and/or other countries.
Sadtler and SearchMaster are trademarks of Bio-Rad Laboratories, Inc. in the United States and/or other countries.
Bruker and OPUS are either trademarks or registered trademarks of Bruker Optics, Inc. in the United States and/or other countries.
All other trademarks are the property of Thermo Fisher Scientific Inc. and its subsidiaries.
OMNIC Specta 2.2 Hotfix 3
-
Build number: 2.2.3.33
-
Release date: January 2026
-
Supported operating systems
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
New Features
None
Resolved Issues
TeIndexer fails to launch when Data Security is used
When Security Suite is installed with OMNIC Specta, the indexer application launches as expected immediately following installation, but will not launch again once closed.
Unable to change directories when opening spectra
When Security Suite is installed with OMNIC Specta, the ability to change directories when opening spectra is removed even when that security policy is disabled.
Printing reports can cause OMNIC Specta to crash when Data Security is used
When Security Suite is installed with OMNIC Specta, printing reports can cause the application to crash if the computer is set to certain language and country combinations.
Using the Peak Area and Peak Height tools causes OMNIC Specta to crash
Due to a missing DLL, attempting to use the Peak Area and Peak Height tools would cause OMNIC Specta to crash.
Known Issues
None
OMNIC Specta 2.2 Hotfix 2
-
Build number: 2.2.2.155
-
Release date: Jan-2023
-
Supported operating systems
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
New Features
None
Resolved Issues
Crash in contaminant search on Windows 11
When you perform a contaminant search on Windows 11, OMNIC Specta crashes.
Known Issues
TeIndexer fails to launch when Data Security is used
When Security Suite is installed with OMNIC Specta, the indexer application launches as expected immediately following installation, but will not launch again once closed.
Unable to change directories when opening spectra
When Security Suite is installed with OMNIC Specta, the ability to change directories when opening spectra is removed even when that security policy is disabled.
Printing reports can cause OMNIC Specta to crash when Data Security is used
When Security Suite is installed with OMNIC Specta, printing reports can cause the application to crash if the computer is set to certain language and country combinations.
- Workaround
-
Change the language and country settings to a supported combination before printing reports.
Using the Peak Area and Peak Height tools causes OMNIC Specta to crash
Due to a missing DLL, attempting to use the Peak Area and Peak Height tools would cause OMNIC Specta to crash.
OMNIC Specta 2.2 Hotfix 1
-
Build number: 2.2.1.51
-
Release date: Apr-2022
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
New Features
None
Resolved Issues
Crashes when performing spectral interpretation
With OMNIC Picta installed, OMNIC Specta program crashes when doing the spectral interpretation and during the multi component search.
Application Name not displayed when running as administrator
If OMNIC Specta was run as administrator, the program name displayed is "MainApplication".
Data Security - When you click on the Open Spectrum in Specta an empty dialog box opens
When using the Open Spectrum icon in the toolbar, no spectra are shown even though it is supposed to show the specified spectra directory.
Known Issues
Crash in contaminant search on Windows 11
When you perform a contaminant search on Windows 11, OMNIC Specta crashes.
Using the Peak Area and Peak Height tools causes OMNIC Specta to crash
Due to a missing DLL, attempting to use the Peak Area and Peak Height tools would cause OMNIC Specta to crash.
OMNIC Specta 2.2
-
Build number: 2.2.0.43
-
Release date: Jun-2017
-
Supported operating systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
-
New Features
Added Data Security (DS) features
-
Add policy to prevent cancel of sample collection
-
Add policy to auto save spectra
-
Add policy to control use of Cancel button on signing dialog
-
Add policy to control prevention of annotation
-
Add audit event for start of a collection
-
Add audit event when cancel login to OMNIC Specta
-
Instrument information is now available for reports
-
The signature dialog will not stop after 3 fails. It will continue to remain open
Resolved Issues
Spectrometer information not saved in header
The spectrum information for some instruments was not included in the spectrum header. This has been resolved.
Library search with updated Hummel library results display incorrect
Spectra from the updated Hummel library were displayed with an incorrect scale. The scale issue has been resolved.
Known Issues
Crashes when performing spectral interpretation
With OMNIC Picta installed, OMNIC Specta program crashes when doing the spectral interpretation and during the multi component search.
Application Name not displayed when running as administrator
If OMNIC Specta was run as administrator, the program name displayed is "MainApplication".
Data Security - When you click on the Open Spectrum in Specta an empty dialog box opens
When using the Open Spectrum icon in the toolbar, no spectra are shown even though it is supposed to show the specified spectra directory.
Using the Peak Area and Peak Height tools causes OMNIC Specta to crash
Due to a missing DLL, attempting to use the Peak Area and Peak Height tools would cause OMNIC Specta to crash.
OMNIC Specta 2.1
-
Build number: 2.1.0.120
-
Release date: Sep-2015
-
Supported operating systems
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit (added in September 2016)
-
New Features
Improved support for importing multiple file types
OMNIC Specta has been revised to import multiple types of spectral files in the spectral file indexer and in File → Open.
Send to OMNIC button disabled
The Send To OMNIC button has been disabled if OMNIC is not installed.
Interaction with the IN10
This has been modified to not interfere with OMNIC or PICTA control of the IN10. The collect feature for this instrument is disabled in Specta.
Microsoft Windows 8.1 is now supported
OMNIC Specta has been tested and is approved for Microsoft Windows 8.1.
Microsoft Windows 10 is now supported
OMNIC Specta has been tested and is approved for Microsoft Windows 10. Added in September 2016.
Resolved Issues
Paper Materials library fully accessible
Some spectra in the library were not accessible. This has been corrected.
Print button in spectral information
Print button in spectral information will now start a print operation.
Known Issues
Report printout may have the wrong paper size or may not fill the paper as intended
Specta → Report → Report Setup → Page Setup and Windows Print → Properties settings may be in conflict. This can result in the wrong paper size or the output not filling the selected paper as expected. To work around this issue, both must be setup for the same page size.
Crashes when performing spectral interpretation
With OMNIC Picta installed, OMNIC Specta program crashes when doing the spectral interpretation and during the multi component search.
OMNIC Specta 2.0
-
Release date: May-2009
-
Supported operating systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
New Features
Raman Data Support
Raman data is now supported with this version of software.
Acquisition (Collect)
Data can now be acquired using a Nicolet iS10 or Nicolet 6700 spectrometer or a Nicolet iZ10 module attached to a Nicolet iS10 or to a Nicolet iN10 microscope. A subset of the OMNIC experimental parameters can be set in OMNIC Specta, while others are set as default to help make collection simpler and easy to use.
Collect Dashboard
The Collect Dashboard can be accessed from the upper toolbar. The Collect Dashboard contains Preview Collect, Sample Collection, Stop Collection, Background Collect and System Status icons to launch Collect services. The dashboard can be accessed easily from anywhere in the application and collection can be finishing as other parts of the application are being used.
Contaminant Search
Use Contaminant Search to identify unknown contaminants in a sample when the major component is known. You can include the entire sample spectrum in the search or just certain spectral regions. You can also specify the maximum number of contaminants to find.
New data displays for Multi-Component Search
Now data can be displayed in Offset, Stack and Terrain view along with Overlay view. Just use the right-click menu to select another display.
Videos for helping you learn and navigate the software
New videos are provided on the Home page for an OMNIC Specta Overview, a Getting Started Video and additional videos to help you understand Manage, Collect, Process, Identify (Overview and Mixture) and Report features.
Step-Me-Through help
Step-by-step directions on how to perform many functions while viewing the software are provided in many areas of the software.
Atmospheric Suppression
Data processing now includes an option to suppress atmospheric artifacts.
Access to other data file formats
File formats listed below can be accessed and used in OMNIC Specta 2.0 software:
-
Digilab® 3200 FT-IR (.dt)
-
Digilab® IMX FT-IR (.dt)
-
Bomem DA3-DA8 FT-IR (.da, .igm, .tra, .ref, .log)
-
Bruker OPUS FT-IR (.0, .1, …, .nnn…)
-
Jasco Spectra Manager (.jws)
-
PerkinElmer® UV Winlab™ (.ig, .sp)
-
Shimadzu® HyperIR FT-IR (.irs)
-
GRAMS (.spc)
Reporting Enhancements
Table sizes and column show/hide functionality are now available features when adding tabular information such as search results to a report.
New color display options
New options are available for selecting color schemes in OMNIC Specta.
Show/Hide Workspace and Result Pane
Showing and hiding the workspace pane and the result pane are now managed from tabs on the right side of the windows instead of using the Up and Down black arrows used in version 1.0.
Multiple Select in Data Tray
The multiple select feature (check mark) in the Data Tray files has now been removed from the software. This functionality was confusing to use and understand.
Sample Information Background display
Background spectra can now be viewed from the Sample Information dialog.
Favorite Names
Favorite names can now be updated after creation.
Favorite Sizes
Adding large numbers of spectra or libraries to a favorite works much better in this version.
Right click options
Additional right click options are available in the software to Send Spectra to be used as references during Subtraction and to use as the contaminant in the Contaminant Search Identify feature.
Multi-Component progress bar
Multi-component search now shows a progress bar.
Library selection display jumping
When you select a library, the display no longer jumps hiding the green "Go" arrow.
Button bar options persist
Button bar options now remain each time the OMNIC Specta application is open.
Baseline correction improvements
Baseline correction has been improved to allow for easier manual and automatic correction.
Exporting data to OMNIC or OMNIC for Dispersive Raman
Data can be exported from OMNIC Specta to OMNIC and OMNIC for Dispersive Raman. OMNIC 8.0 or higher will work for sending spectra back and forth between applications and OMNIC for Dispersive Raman 8.1 or higher will allow for sending Raman spectra back and forth between the applications. OMNIC and OMNIC for Dispersive Raman software versions prior to 8.0 and 8.1 respectively do not support this option.
Resolved Issues
Adobe Acrobat 9.0 is supported
Adobe® Acrobat® 9.0 is now supported in OMNIC Specta version 2.0.
Search spectra skip feature
If spectra in a library or favorite are unchecked to skip in searching, they are now removed from the search results. In the previous version these spectra could still be reported in the search hit list.
Color options
Table sizes and column show/hide functionality are now available features when adding tabular information such as search results to a report.
Knowledge Base usage without Adobe Acrobat installed
An error is now returned when using the Knowledge Base feature without having installed Adobe Reader.
OMNIC Specta DS
Specta DS now shows spectra in Open Dialog from OMNIC 8 DS.
Multiple report pages
Multiple pages in report now show correct page numbers instead of just 1 of 1.
Search results X-Axis display range
Changing search results resets X-axis display range, now the X-axis display range stays zoomed as specified by the user.
Hide Report item preview
The Report item preview is hidden once the item is added to the report.
Interpretation Guide window
When extracting pages from the Interpretation Guide no longer causes the interpretation window to close.
Page numbering when extracting and adding pages
The page number now updates correctly when pages are added and extracted from the current report.
OMNIC Result matching
Identify Spectral Search results now match the results reported in OMNIC.
Data Tray
Only one version of each spectrum can be accessible from the Data Tray.
Localization
All icons and options are now localizable.
Multiple search hits are the same
Multiple search results containing the same material will no longer be displayed in the search results. Each material will only be displayed a single time.
Right click option to removed Baseline Correct points
A right click option is now available to removed points from the baseline.
Interpret display in Absorbance
Interpret display now shows all green blocks in Absorbance and %Transmittance.
My Data "Folders on Disk"
My Data "Folders on Disk" now updates when a spectrum is deleted.
Searching using .spg files
Multi-component search now returns the correct match values when using an spg file as a reference.
Signing User Libraries
To sign user libraries please use the OMNIC application features in Library Manager.
Known Issues
Data Execution Prevention message
Windows Notification: "Data Execution Prevention has closed TeIndexer"
There is a known issue with the Microsoft Windows "Data Execution Prevention" feature on Vista which can cause this message to appear.
The message is meaningless and can be ignored. Contrary to the message the TeIndexer application will continue to run and function normally. Microsoft is aware of this problem in Vista; refer to article KB948468 for details.
Jasco and Bruker spectral data files
Some older versions of Jasco and Bruker spectra are not recognized by Specta.
There is a known issue with some older versions of Jasco and Bruker spectrum files that prevents Specta from using these files.
Send spectra from OMNIC Specta to OMNIC for Dispersive Raman and Send spectra from OMNIC for Dispersive Raman to OMNIC Specta
This feature will require OMNIC for Dispersive Raman version 8.1 or higher.
Old third-party spectrum formats are no longer readable in Windows 7 64-bit
Some of the older format readers do not function on Windows 7 64-bit systems. When the Specta indexer encounters this situation, Windows displays an "Unsupported 16-bit Application" message. Until new readers are available, the old spectrum files should be moved to a folder the Specta indexer will not attempt to read.
Crashes when performing spectral interpretation
With OMNIC Picta installed, OMNIC Specta program crashes when doing the spectral interpretation and during the multi component search.
OMNIC Specta 1.0
-
Release date: Dec-2008
-
Supported operating systems
-
Windows XP SP3 32-bit
-
Windows 7 32-Bit
-
Windows 7 64-bit
-
New Features
Initial Release, all features are new
Resolved Issues
Initial release, there are no resolved Issues
Known Issues
None
Attachment(s):
| File | Last Modified |
|---|---|
| OMNIC Specta Release Notes.pdf | January 09, 2026 |