OMNIC SST Release Notes
OMNIC SST Release Notes
About This Document
This document contains a revision history of OMNIC SST, including new features that may not be included in the User’s Guide, resolved issues, and known issues.
Trademarks
Microsoft, Internet Explorer, and Windows are either trademarks or registered trademarks of Microsoft Corporation in the United States and/or other countries.
ToolBook is either a trademark or registered trademark of SumTotal in the United States and/or other countries.
Semprex is either a trademark or registered trademark of Semprex Corporation in the United States and/or other countries.
PerkinElmer is either a trademark or registered trademark of PerkinElmer, Inc. in the United States and/or other countries.
Sadtler and SearchMaster are trademarks of Bio-Rad Laboratories, Inc. in the United States and/or other countries.
Bruker and OPUS are either trademarks or registered trademarks of Bruker Optics, Inc. in the United States and/or other countries.
All other trademarks are the property of Thermo Fisher Scientific Inc. and its subsidiaries.
OMNIC SST 9.16 Hotfix 5
-
Build number: 9.16.5.240
-
Release date: August 2026
-
Supported operating systems
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
|
Note
|
There were no changes to OMNIC SST in this release, but, as of 9.14, it is always rebuilt and released with the OMNIC software. |
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.16 Hotfix 4
-
Build number: 9.16.4.233
-
Release date: February 2026
-
Supported operating systems
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
|
Note
|
There were no changes to OMNIC SST in this release, but, as of 9.14, it is always rebuilt and released with the OMNIC software. |
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.16 Hotfix 3
-
Build number: 9.16.3.244
-
Release date: January 2026
-
Supported operating systems
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
|
Note
|
There were no changes to OMNIC SST in this release, but, as of 9.14, it is always rebuilt and released with the OMNIC software. |
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.16 Hotfix 2
-
Build number: 9.16.2.213
-
Release date: December 2025
-
Supported operating systems
-
Windows 10 64-bit Version 1607 and newer
-
Windows 11 64-bit
-
|
Note
|
There were no changes to OMNIC SST in this release, but, as of 9.14, it is always rebuilt and released with the OMNIC software. |
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.16 Hotfix 1
-
Build number: 9.16.1.188
-
Release date: March 2025
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
| There were no changes to OMNIC SST in this release, but, as of 9.14, it is always rebuilt and released with the OMNIC software. |
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.16
-
Build number: 9.16.0.159
-
Release date: September 2024
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
| There were no changes to OMNIC SST in this release, but, as of 9.14, it is always rebuilt and released with the OMNIC software. |
OMNIC SST 9.15
-
Build number: 9.15.0.119
-
Release date: May 2024
-
Supported operating systems
-
Windows 10 64-bit
-
Windows 11 64-bit
-
| There were no changes to OMNIC SST in this release, but, as of 9.14, it is always rebuilt and released with the OMNIC software. |
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.14
Release Date: Jan-2024
Supported Operating Systems
-
Windows 10 64-bit
-
Windows 11 64-bit
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.13 Hotfix 5
Release Date: Jun-2023
Supported Operating Systems
-
Windows 10 64-bit
-
Windows 11 64-bit
New Features
None
Resolved Issues
OMNIC DDE Step-Scan Phase Calculation is wrong at low frequencies
At SST frequencies of 5-50Hz, the DDE version of the phase calculation is consistently wrong, resulting in a quadrature interferogram that is larger than the reference interferogram.
Known Issues
None
OMNIC SST 9.9
Release Date: Feb-2019
Supported Operating Systems
-
Windows 7 32-bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
None
Resolved Issues
Collection issue
The collections would proceed but never finish.
Known Issues
None
OMNIC SST 9.8 Hotfix 1
Release Date: Oct-2017
Supported Operating Systems
-
Windows 7 32-bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
None
Resolved Issues
Unhandled error message when collecting with dual channels
An unhandled error message can occur after the collection has completed and the data is being added to a window.
Known Issues
None
OMNIC SST 9.8
Release Date: May-2017
Supported Operating Systems
-
Windows 7 32-bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
Added support for GaGe 1442e PCIExpress card
The GaGe 1442e PCIExpress case can used as a replacement for the GaGe 1442 PCI card.
Resolved Issues
None
Known Issues
None
OMNIC SST 9.7
Release Date: Sep-2016
Supported Operating Systems
-
Windows 7 32-bit
-
Windows 7 64-bit
-
Windows 8.1 64-bit
-
Windows 10 64-bit
New Features
Added support for Windows 10.
Product now can be used on Windows 10
Resolved Issues
None
Known Issues
None
OMNIC SST 9.6
New Features
None
Resolved Issues
Failure in iS50R TRS data collect when the data collect is paused
Pressing "Stop" rather than "Resume" after pausing the TRS data collect, causes an OMNIC crash.
Incorrect spectrum with De Haseth phase correction
The result of the De Haseth phase correction is not correct; some of the results are inverted.
Amplitude in AM is much less than the actual signal at the channel input
The problem was fixed by changing the setting for the high pass filter to 1.
Incorrect filter settings for step scan experiments
Rather than setting the filters for the linear scan Experiment Setup menu, they will be set automatically to match the experiment selection from the SST menus.
Known Issues
None
OMNIC SST 9.5
No changes for this release except the version number.
New Features
None
Resolved Issues
None
Known Issues
None
OMNIC SST 9.4
New Features
Added support for Windows 8.1.
Resolved Issues
iS50R TRS Gage series data causes exception
Reprocessing a series file with 100 or so spectra collected using a TRS Gage card causes an exception to occur.
Memory leak in 8700 Dual channel collect
A memory leak using the dual channel collect over long periods of time was detected.
Known Issues
None
OMNIC SST 9.1 Hotfix 3
New Features
None
Resolved Issues
SST Menu items not enabled on Dual Channel only systems
On systems that only support the Dual Channel measurements, there were situations where the SST menu items for performing the measurements would not get enabled.
| The earlier OMNIC 9 Hotfixes did not affect SST so no entries were made in the release notes. |
Known Issues
None
OMNIC SST 9.0
New Features
Support for new iS50R model.
Resolved Issues
None
Known Issues
None
OMNIC SST 8.3
New Features
None
Resolved Issues
Post Trigger Delay Time inaccurate with GaGe 1422
A problem with the post trigger delay in time resolve measurements being inaccurate has been fixed. The actual delay was often double the delay entered in the setup dialog.
Known Issues
None
OMNIC SST 8.1
New Features
Support for new 8700 model.
Resolved Issues
None
Known Issues
None
OMNIC SST 7.4
New Features
None
Resolved Issues
Zero filling when step-scan spectra are Reprocessed
In some cases an additional level of zero filling would be applied when a step-scan spectrum is reprocessed.
Known Issues
None
OMNIC SST 7.3 Service Pack 1
New Features
Support of the Nicolet 8700 (USB version)
Support has been added for the USB version of the Nicolet 8700 spectrometer. This includes the extension of the PM modulation amplitude to 4.5 lambdas and three enhancements of step-scan time-resolved data collection: The optional use of an additional input (Sync), a shorter (1 microsecond) minimum sampling interval when using the spectrometer digitizer, and the ability to use multiple triggers per step when using the spectrometer digitizer.
Resolved Issues
None
Known Issues
OMNIC Data Security
If OMNIC Data Security is installed on your system, the "Require Signature When Saving Spectrum" policy should be disabled when doing a PEM Kinetics data collection so that the collections will not be interrupted by requests for digital signatures
OMNIC SST 7.3
New Features
Support of the Nicolet 8700
Support has been added for the Nicolet 8700 spectrometer. All of the capabilities of the Nexus 870 spectrometer are retained in this spectrometer.
Resolved Issues
None
Known Issues
None
OMNIC SST 7.2
New Features
New controls in Step-Scan Time Resolved setup dialog
A checkbox labeled Stop trigger signal after collect and an edit control labeled AC/DC amplification factor have been added to the Step-Scan Time Resolved setup dialog.
Advanced Applications command
A new command named Advanced Applications has been added to the SST menu. It provides access to a PDF-format copy of the book "Advanced FT-IR Spectroscopy" by respected scientist, Dr. Eric Y. Jiang, Ph.D.
Resolved Issues
Gain and Filter controls in setup dialogs
The Gain and Filter controls in the Dual Channel Polarization Modulation and Step-Scan Multiple Modulation setup dialogs are hidden unless the spectrometer type is a Magna-IR 850 or 860. These features are not available on other spectrometers.
Known Issues
None
OMNIC SST 7.1
New Features
Support of the dual channel Nicolet 6700
Support has been added for the dual channel option of the Nicolet 6700.
Resolved Issues
None
Known Issues
None
OMNIC SST 7.0
New Features
Larger sample spacing in step-scan experiments
Larger sample spacing values may be selected in the setup dialogs for step-scan experiments. The largest value allowed will depend on the spectral range specified in the Bench tab of Experiment Setup.
Trigger stepping with SYNC
An additional control labeled Trigger stepping with SYNC is available in the Step-Scan Amplitude and Phase Modulation and Step-Scan Time Resolved experiment setup dialogs. (This is available only on Nexus 870 systems with Flash code version 7.16 or higher.)
OMNIC SST Installation Directories
When you install OMNIC SST, the folders into which its files are copied depend on where OMNIC has been installed on your system. The default folders are C:\My Documents\OMNIC for data files, C:\My Documents\OMNIC\pdf for documentation files and C:\Program Files\OMNIC for program (executable) files and Help files.
OMNIC SST Files
The OMNIC SST install program installed the following files in the Program Files\OMNIC directory on your hard disk drive:
-
OMNIC_vi.dll
-
gage_drv.dll
-
sst_enu.chm
-
magnadia.exe
-
def850.cfg
-
spectrometer-specific hardware help files
AdvFT_IRSpec.pdf is installed in the Program Files\OMNIC\pdf directory.
In addition, several experiment files were installed in the My Documents\OMNIC\Param directory on your hard disk. These can be used to configure the software for various step-scan and dual-channel linear scan experiments. They are listed and described briefly below.
| File Name | Description |
|---|---|
|
860am.exp |
Default Experiment file for Amplitude Modulation Step Scan Experiments |
|
860pm.exp |
Default Experiment file for Phase Modulation Step Scan experiments |
|
mcta4rap.exp |
Default Experiment file for Rapid Scan Data Collection at 4 cm-1 resolution |
|
pas1slow.exp |
Default Experiment File for Linear Scan Photoacoustic Spectroscopy at 0.0633 cm/sec mirror velocity |
|
pas2slow.exp |
Default Experiment File for Linear Scan Photoacoustic Spectroscopy at 0.0158 cm/sec mirror velocity |
|
pas_stsc.exp |
Default Experiment File for Phase Modulation Step Scan Photoacoustic Spectroscopy Using 400 Hz Phase Modulation |
|
siqussp0.exp |
Default Experiment File for Extended Spectral Range Measurements to 25,000 cm-1. Requires Quartz Beam splitter and Silicon Detector |
|
siqussp1.exp |
Default Experiment File for Extended Spectral Range Measurements to 15,000 cm-1. Requires Quartz Beam splitter and Silicon Detector |
|
IRRAS_R.exp |
Default Experiment file for Dual Channel Polarization Modulation IRRAS experiments (Right Side TOM configuration) |
|
IRRAS_L.exp |
Default Experiment file for Dual Channel Polarization Modulation IRRAS experiments (Left Side TOM configuration) |
|
VCD_R.exp |
Default Experiment File for Dual Channel Polarization Modulation VCD experiments (Right Side TOM configuration) |
|
VCD_L.exp |
Default Experiment File for Dual Channel Polarization Modulation VCD experiments (Left Side TOM configuration) |
|
VLD.exp |
Default Experiment File for Vibrational Linear Dichroism using Main Spectrometer MCT-A Detector |
How to Contact Us
Current contact information is located at https://www.thermofisher.com
Select the "Contact Us" icon at the top of the screen
Attachment(s):
| File | Last Modified |
|---|---|
| OMNIC SST Release Notes.pdf | August 26, 2026 |