Raman Autosampler Release Notes
About This Document
This document contains a revision history of Raman Autosampler, including new features that may not be included in the User’s Guide, resolved issues, and known issues.
Trademarks
Microsoft, Internet Explorer, and Windows are either trademarks or registered trademarks of Microsoft Corporation in the United States and/or other countries.
ToolBook is either a trademark or registered trademark of SumTotal in the United States and/or other countries.
Semprex is either a trademark or registered trademark of Semprex Corporation in the United States and/or other countries.
PerkinElmer is either a trademark or registered trademark of PerkinElmer, Inc. in the United States and/or other countries.
Sadtler and SearchMaster are trademarks of Bio-Rad Laboratories, Inc. in the United States and/or other countries.
Bruker and OPUS are either trademarks or registered trademarks of Bruker Optics, Inc. in the United States and/or other countries.
All other trademarks are the property of Thermo Fisher Scientific Inc. and its subsidiaries.
Raman Autosampler 6.0
Release Date: Nov-2023
Significant Changes
This version of the Raman Autosampler application is a complete rewrite of the previous version. As such, some of the legacy functionality has been removed as listed below:
-
Support for OMNIC Macros/Basic Has Been Removed
-
OMNIC Menu Items Removed
-
Custom Column Labels Are No Longer Supported
-
Support for .ras files has been removed
New Features
None
Resolved Issues
Unable to Specify Spectrum Title or File Name
All spectra were assigned a timestamp title and automatically generated file name.
Known Issues
None
Raman Autosampler 5.1
New Features
Added Silent Mode
Added SilentOn and SilentOff commands to the Raman Autosampler command line to suppress completion messages when using OMNIC Macros\Basic.
Removed Motor Hysteresis Compensation for Raman Spectrometers
The motor hysteresis compensation feature was removed for Raman spectrometers. It is automatically handled by OMNIC for Dispersive Raman.
Removed Automatic Reset Wheel on Startup
A command line option has been added for the Raman Autosampler application to automatically reset the wheel.
Resolved Issues
Out of Bounds Error When Spreadsheet is Empty
An out of bounds error could occur when the spreadsheet was empty.
Spin Function Could Fail on FT-Raman Systems
Firmware limitations for the motor control could cause the spin function to fail on FT-Raman systems. The spin function now automatically reverses direction when it approaches the limit.
System Could Time Out Waiting for OMNIC Macros\Basic to Load
The timeouts for the Raman Autosampler application have been increased to allow OMNIC Macros\Basic to load.
The Adjust Wizard Did Not Work Correctly on Almega Spectrometers
Changes to OMNIC for Dispersive Raman resulted in the Adjust wizard not working correctly on Almega spectrometers. The Adjust wizard now works correctly on Almega spectrometers.
Known Issues
None
Raman Autosampler 5.0
New Features
Added Support for DXR
Added support for DXR Raman spectrometers.
Resolved Issues
None
Known Issues
None
Raman Autosampler 4.0
New Features
Added Support for MoveTo from Bench Tab in OMNIC for Dispersive Raman
Added support for the MoveTo button on the Bench tab in OMNIC for Dispersive Raman. This button is used to move the Raman motorized stage to the position specified in the text box. The Raman Autosampler application must be running for this feature to work.
Resolved Issues
None
Known Issues
None
Raman Autosampler 3.1
New Features
New OMNIC Macros\Basic tasks
Added tasks to OMNIC Macros\Basic which support the remote operation features of Raman Autosampler. These new tasks appear near the bottom of the Collect section in the OMNIC Macros\Basic tasks list.
The new tasks are:
| Task | Description |
|---|---|
|
RAS Display Spreadsheet |
Shows the Raman autosampler spreadsheet |
|
RAS Reset Wheel |
Resets the Raman autosampler sample wheel and leaves it at position #1 |
|
RAS Move To Position |
Moves the Raman autosampler wheel to the position you specify |
|
RAS Exit |
Closes the Raman autosampler application |
These commands are described in the document Raman Carousel Autosampler User’s Guide - Appendix.pdf located on the software CD.
Resolved Issues
None
Known Issues
None
Raman Autosampler 3.0
New Features
Remote Operation
Added support for remote operation with or without the spreadsheet graphical user interface. These commands are described in the document Raman Carousel Autosampler User’s Guide - Appendix.pdf located on the software CD.
OMNIC Menu Commands
Added Raman Autosampler commands to the OMNIC Collect menu. These commands include:
-
Display Spreadsheet
-
Reset Wheel
-
Move to Position
-
Move to Next Allowed Position
-
Spin Sample
-
Exit
Auto start Raman Autosampler with OMNIC
Added capability to automatically start the Raman Autosampler application when OMNIC starts. This capability is described in the document Raman Carousel Autosampler User’s Guide - Appendix.pdf located on the software CD.
Resolved Issues
A macro error message followed by application error
When you click Start to begin the analysis of samples entered in the spreadsheet, the analysis macro for the first sample begins to run. An error message occurs followed by an application error which closes the Nicolet Raman Autosampler application.
Known Issues
None
Raman Autosampler 2.5
New Features
None
Resolved Issues
Issue with N2 Bench
No other information is available.
Known Issues
None
Raman Autosampler 2.4
New Features
Added Support for up to 100 Different Sample Wheel Types
Updated INI file reader and UI code to support up to 100 differnt sample wheel types.
Resolved Issues
None
Known Issues
None
Raman Autosampler 2.3
There do not appear to be any official release notes for this version.
Raman Autosampler 2.2
There do not appear to be any official release notes for this version.
Raman Autosampler 2.1
There do not appear to be any official release notes for this version.
Raman Autosampler 2.0
New Features
Macro Shows Only File Name in Spreadsheet
The directory path for macro files is no longer displayed in the Macro column cells; only the file name. The directory path is saved, however, in a hidden cell associated with each macro. Each macro may have its own unique path.
| This change means that existing .RAS files from 1.0 cannot be opened in 2.0. You must recreate them in 2.0. |
Macro File Selection Defaults to OMNIC Macro Directory
When you click the Browse button to select a macro, the File Open dialog is displayed pointing to the directory where the macro is located. If the macro cell is blank, the File Open dialog opens in the directory which was last used to select a macro. If the application has just been started and this is the first macro selection, the dialog opens pointing to the macro directory path specified in OMNIC.
Resolved Issues
Stopping a Macro Does Not Stop the Autosampler
The application has been changed so that stopping a macro before it is finished will also stop the autosampler.
Raman Autosampler Loses Track of Position When Experiment Setup Task is Run
OMNIC Macros\Basic has been changed so that the Experiment Setup task no longer repositions the Raman motorized stage. This was causing the Raman autosampler to lose track of its position.
Fill Series Fails for Single Digit Values
A bug in Fill Series has been fixed so that it now works correctly with single digits.
Adding Columns to the Spreadsheet Causes Fatal Error
A bug with adding columns to the spreadsheet then trying to resize the added column has been fixed. It no longer causes a fatal error.
Sample Wheel Position Could Become Inaccurate After Repeated Runs
The application has been changed so that the wheel is driven against the hard stop prior to beginning each run. This can be quite annoying but it is the only way to guarantee that the wheel is in known position during the run.
Known Issues
None
Raman Autosampler 1.0
There do not appear to be any official release notes for this version.
How to Contact Us
Current contact information is located at https://www.thermofisher.com
Select the "Contact Us" icon at the top of the screen
Attachment(s):
| File | Last Modified |
|---|---|
| Raman Autosampler.pdf | November 27, 2023 |