Empower OQ - Processing DAD WLA Scan Test
Empower OQ - Wavelength Accuracy Scan Report not showing data
Issue
The DAD Wavelength Accuracy test (using caffeine) can be accomplished with a single injection using 3D Scan capabilities in SII for Empower. A wavelength range (190-350 nm) of caffeine can be scanned so that the 2 maxima (205 nm, 273nm) and minima (245 nm) can be extracted via processing, and then reported via Empower.
Environment
- Empower 3
- SII 1.1 or later
- OQ
- DAD
- WLA Scan
Resolution
The following video demonstrates the process for:
- Opening the scanned channel collected from acquisition
- Applying the 3D Scan Method Set / Processing Method to the data
- Reviewing the extracted spectrum and Caffeine test results
- Processing the channel to commit the 3D integration
- Creating the final test report
- For Troubleshooting the report see: Empower OQ - Wavelength Accuracy Scan Report not showing data