200409TP - Rev 2 - Waters BUS SAT_IN Module Installation Guide
Install the Waters Bus SAT/IN module to digitize up to two detector channels and send data to a Bus LAC/E card via the I/O Distribution Box. Select an appropriate site, then connect serial, analog, and event cables; add optional BCD; attach the power supply. Power up devices, verify LEDs, and configure ports and methods in Millennium. Observe cable length limits, ground shields, and disconnect power before servicing. Use specs, spares, and warranty as needed.
Description:
The Waters Bus SAT/IN Module Installation Guide contains information needed to install a Waters™ Bus SAT/IN™ module. The Bus SAT/IN module translates signals from analog devices to digital form, and transmits those signals to the Waters Bus LAC/E card inside a host personal computer.
This installation guide is intended for anyone who must install a Bus SAT/IN module. This guide assumes that the Waters Bus LAC/E™ card is installed in the host computer and working properly.
Environment:
- Chromeleon
- Waters Driver
Attachment(s):
| File | Last Modified |
|---|---|
| 200409TP - Rev 2 - Waters BUS SAT_IN Module Installation Guide.pdf | January 20, 2022 |