Thermo Fisher Scientific
Agilent Drivers for Chromeleon 2.3
This package supplies drivers that allow Chromeleon 7 to control Agilent chromatography instruments via the Agilent instrument control framework. It provides a list of supported instruments and firmware versions, along with detailed documentation and step-by-step installation guidance. A readme file offers remarks and updates. The distribution also contains software licensing terms for review and compliance. Review all guides before install to confirm set up compatibility with confidence.
Description: 
A new set of drivers to control Agilent LC and GC hardware. New features include:
  • Dual Simultaneous Injection for GCs
  • Dynamic Licensing (see Change Detail below)
  • Adding lines to running sequences
  • Separate user guides for GC and LC
Please see the Release Notes provided with the drivers for more new features and further details on these features.
Environment:
  • Chromeleon 7.2 SR5 (including all MUs)
  • Chromeleon 7.2.10 (including all MUs)
  • Chromeleon 7.3
  • Was this article helpful?