Thermo Fisher Scientific
Agilent Drivers for Chromeleon 2.2 Update 1
This package supplies drivers that allow Chromeleon 7 to control Agilent chromatography instruments via the Agilent instrument control framework. It provides a list of supported instruments and firmware versions, along with detailed documentation and step-by-step installation guidance. A readme file offers remarks and updates. The distribution also contains software licensing terms for review and compliance. Review all guides before install to confirm set up compatibility with confidence.
Description: 
The following Agilent components are included in Agilent Drivers for Chromeleon 7 (Rev. 2.2 Update 1):
  • Agilent Instrument Control Framework 2.6 Update 1 
  • Agilent LC Drivers A.02.19 SR2 
  • Agilent ELSD Drivers A.01.07 DU1 
  • Agilent GC-HS Drivers 3.0 SR4
Environment:
  • Agilent ICF
  • Chromeleon 7
  • Was this article helpful?