Error 1172
Issue
The following error appears when submitting an instrument method in Tracefinder:
Environment
- LCMS
- TraceFinder
Resolution
If no error is seen using Xcalibur, TraceFinder Configuration should be checked.
Confirm that the Multichannelling option is selected under “optional features”. Select channels available. Please note
that “Intelligent sequencing” feature will not be available and that “Full sequence submission” will be automatically
selected.
Attachment(s)
| File | Last Modified |
|---|---|
| error 1172 - image 02.png | January 10, 2022 |
| error 1172 - image 01.png | January 10, 2022 |