Thermo Fisher Scientific
Nicolet X700 Everglo MIR Source Failure Mode
Issue
As a Everglo MIR source ages, the output reduces.
Environment
  • Nicolet X700
  • Everglo MIR source
Analysis
This reduction is due to two factors, the impedance in the element increases allowing less current throught the element along with the hot spot of the element shifts from the center to the edge.
Energy reduction due to the shift in the hot spot of the element will be amplified when the aperture is reduced in size.
  • Was this article helpful?