How to change location of standards and samples in iCE 3000 Furnace auto-sampler rack.
Issue
Customize standards or samples positions in Furnace auto-sampler rack.
Environment
- iCE3400
- iCE3500
- S Series
- M6.
Resolution
In the Solaar software, open Method, click on Sequence tab, click on ASLG (Auto-sampler Loading Guide). Click on any standard or sample from rack and drag then drop into any position.
Attachment(s)
File | Last Modified |
---|---|
ASLG.jpg | January 12, 2023 |