Thermo Fisher Scientific
Radial POP Interface Kit for Duo
Description: 
The dual view purged optical path plasma interfaces of the Thermo Scientific™ iCAP™ 7000 Plus Series ICP-OES enhance long term stability and improve detection limits
in the wavelength region below 180 nm for both axial and radial measurements. This Technical Note: SP-43438-ICP-OES-Purged optical path plasma-iCAP 7000 Plus-SP43438-EN.pdf  provides further details on this new interface for the radial view.
The P/N for the Radial POP Interface Kit is: BRE0011655. It contains the following items:
1x POP interface
2x glass window
2x O-ring
1x alignment tool
Radial POP Interface kit.png
Environment:
  • iCAP 7000 Series
  • Was this article helpful?