Chiller leaks
Issue:
Chiller leaks
Environment:
- TF 900/2500/1400
Resolution:
Found Stainless mesh had fallen into the reservoir, allowing effluent to spill over top of opening and running down inside of chiller cabinet.
Chiller leaks
Found Stainless mesh had fallen into the reservoir, allowing effluent to spill over top of opening and running down inside of chiller cabinet.