Error 8 - Calibration needs to be saved as a factory reset
Issue
Brand new chiller out-of-the-box displaying "Er 8" which indicates that Calibration needs to be saved as a factory reset
Environment
- ThermoFlex 900
- ThermoFlex 2500
Resolution
Enter the Service Function:
1. Press and hold <enter> and then press <mode> three times in a row
2. The controller will beep. If not, redo holding <Enter> and then press <mode> 3 times in a row
Enter the calibration loop:
1. Press <mode> to display SP
2. Press <mode> again to display Setup
Note: You have 60 seconds to press <mode> or the Service Function will time out
3. Press <mode> until you see diA9 displayed
4. Press and hold the <Up arrow> followed by pressing <mode> again to enter the Calibration Loop
Reset sensor to factory value:
With rtd1 displayed, press and hold the <Up arrow> key and then press the <Down arrow> key until the display flashes between gEt and rESEt and then press the <Enter> key
The unit will beep and the display will go blank to indicate the calibration data was retrieved from the RESET location in memory
Analysis
Original error code was "Er 6" - Calibration, maintenance counters and setup variables were reset. Once this error was cleared, Er 8 appeared.