Not Cooling Down to Set Point
Issue
The chiller is not cooling down to set point
Environment
ThermoFlex 900
ThermoFlex 1400
ThermoFlex 2500
Resolution
There have been cases mainly on the ThermoFlex 900 chillers where the software code has been configured incorrectly for the Unit code. Use this procedure to check if the software code has been configured with the code that is specific for the chiller: ThermoFlex Basic Re-Configuration - 16 digit.pptx
Attachment(s)
File | Last Modified |
---|---|
ThermoFlex Basic Re-Configuration - 16 digit.pptx | April 26, 2023 |