Thermoflex Chiller HPC error
Issue
The Thermoflex 900 chiller turns off due to a HPC error caused by the Fan Motor siezing
Environment
Thermoflex 900, Thermoflex 1400, Thermoflex 2500 chillers
Resolution
- Turn off the chiller and unplug the power cord.
- Open the right side cover (Figure 1).
(Figure 1)
3. Clean any dust with a vacuum at the air cooling fan and air cooling heat exchanger.
4. Add lubricant (WD-40) to the axial of the air cooling motor and rotate the fan using anything that can be used to turn the fan through the fan guard ex: pencil (Figure 2).
(Figure 2)
5. If the error persists then replace the fan motor (see instructions in attachments).
Analysis
The Thermoflex chiller displays a HPC error.
Cause
The fan motor has seized causing a temperature increase in the cooling liquid.
Attachment(s)
File | Last Modified |
---|---|
P1.jpg | January 11, 2023 |
ThermoFlex 900 Fan Motor Replacement.pdf | November 16, 2022 |