Thermo Fisher Scientific
Audit Trail not showing in SOLAAR Security
If in SOLAAR Security Admin, the System Log appears blank, open the Administer SOLAAR Security Options as a PC administrator. Alternatively, refer to page 4-1 of Rev B - BRE0017280 - SOLAARsecurity Software Manual for detailed steps.
Issue
The System Log in SOLAAR Security Admin is blank
Environment
  • SOLAAR Security
Resolution
  • Open the Administer SOLAAR Security Options as a PC administrator
An Alternate approach to the steps listed in the video above, is listed on page 4-1 of  Rev B - BRE0017280 - SOLAARsecurity Software Manual
  • Was this article helpful?