Configuring the Triplus RSH / 100 LS Autosampler to use 2 or 10 mL
Issue
Configuring the Triplus RSH / 100 LS Autosampler to use 2 or 10 mL - CV Magnetic Sample Vials for Headspace / SPME Analysis
Environment
- Triplus RSH
- Headspace
- SPME analysis
Resolution
- Switch the Agitator’s Vial Type Position from 20-CV Magnetic to 10-CV Magnetic in the Extended User Level Administrator Mode of Virtual Terminal.
- Switch the Trayholder's Tray Vial Type from 20-CV Magnetic to 10-CV Magnetic in the Extended User Level Administrator Mode of Virtual Terminal.
- Install the 6 - 2 or 10 mL Agitator spacers into the 6 heated Agitator vial position spots.
The Spacer PNs:
- TriPlus RSH - Agitator Adapter for 2mL Vial (Set of 6) P/N1R77010-1067
- TriPlus RSH - Agitator Adapter for 10mL Vial (Set of 6) P/N1R77010-1068