Thermo Fisher Scientific
Heater that is not heating
If the SSL heater in your Trace 1300 Series GC is not heating up, it is recommended to replace it. The part number for the GCMS Trace 1300 Series SSL heater is 22206154. Ensure you acquire this replacement part to resolve the heating issue.
Issue
I have a SSL heater that is not heating up.
Environment
Trace 1300 Series
Resolution
Recommended to replace, GCMS Trace 1300 Series part number is 22206154.
  • Was this article helpful?