903061 - Rev C - Next Generation Control Board - Product Guide
The Next Generation Control Board Product Guide details the control board used in pumps, including its construction, user interface, and communication capabilities. It explains the setup menu, keypad functions, LED indicators, serial and USB communication, firmware updates, and motor drive board connection.
Description:
This document describes in detail the Control Board that is supplied with Next Generation Pumps and Pump Kits. The Control Board provides user-interface capabilities for the Motor Drive Board, SSI pumps and pump kits, including communications and front-panel display.
Environment:
- AXP Pump
- Next Generation Control Board
Attachment(s):
| File | Last Modified |
|---|---|
| 903061 - Rev C - Next Generation Control Board - Product Guide.pdf | January 07, 2022 |