Thermo Fisher Scientific
903062 - Rev A - External Control for Next Generation SSI Pumps
The Next Generation Control Board Product Guide details the control board's user interface, communication capabilities, and construction for Next Generation pumps. It covers setup menus, keypad functions, LED indicators, serial and USB communication, firmware updates, motor drive board connections, and power-up functions.
Description: 
This document describes in detail the External Control Board that is frequently supplied with Next Generation Pumps and Pump Kits. The External Control Board provides external pump control (run, stop, and voltage), output monitoring (fault, relay) and fan power for a wide assortment of pumps from Scientific Systems, Inc.
Environment:
  • AXP Pump
  • SSI Pumps
  • External Control Board
  • Was this article helpful?