Thermo Fisher Scientific
903065 - Rev B - Next Generation Motor Drive Board - Product Guide
The Next Generation Motor Drive Board Product Guide details the motor drive board used in Next Generation pumps, including its construction, communication interfaces, and control features. It covers serial communication, pressure transducer connections, motor flag operation, power input, motor connections, external control, and Modbus communication protocols.
Description: 
This document describes in detail the Motor Drive Board that is supplied with Next Generation Pumps and Pump Kits. The Motor Drive Board provides motor drive and pump control for a wide assortment of standalone pumps and pump kits from Scientific Systems, Inc.
Environment:
  • AXP Pump
  • Was this article helpful?