031928 - Rev 07 - Preventive Maintenance Procedure Dionex Aquion and ICS-1000, ICS-1100, ICS-1500, ICS-1600, ICS-2000, ICS-2100
The document outlines the preventive maintenance procedure for various Dionex Ion Chromatography Systems, including models Aquion, ICS-1000, ICS-1100, ICS-1500, ICS-1600, ICS-2000, and ICS-2100. The procedure includes rebuilding the high-pressure injection valve, auxiliary valve, replacing pump check valve cartridges and seals, waste valve and priming valve O-rings, and end-line filter. The maintenance should be performed annually and before Performance Qualification tests.
Description:
This manual describes a preventive maintenance procedure for the Thermo Scientific Dionex Aquion, ICS-1000, ICS-1100, ICS-1500, ICS-1600, ICS-2000, and ICS-2100 Ion Chromatography Systems.
Environment:
- Aquion
- ICS-1000
- ICS-1100
- ICS-1500
- ICS-1600
- ICS-2000
- ICS-2100
Attachment(s):
| File | Last Modified |
|---|---|
| 031928 - Rev 07 - Preventive Maintenance Procedure Dionex Aquion and ICS-1000, ICS-1100, ICS-1500, ICS-1600, ICS-2000, ICS-2100 | December 30, 2021 |