Skip to main content
Environment:
- Aquion
- Easion
- Integrion
- ICS-1X00, ICS-2X00
- ICS-3000
- ICS-4000
- ICS-5000
- ICS-5000 Plus
- ICS-6000
Pump Head Assemblies:
Primary pump heads:
- 067701 – ASSY, HEAD, PRIMARY, ICS900 for Easion.
- 057937 – HEAD ASSY, PRIMARY, SERIES PK for Aquion, ICS-1X00, ICS-2X00.
- 062053 - (DISCONTINUED) HEAD ASSY,PRIMARY,PMP,ICS-3000 for ICS-3000/5000 analytical pumps is no longer an available part and cannot be ordered. Please see PSB-ICSP-1117 - ICS-5000 DP and SP Pumps Spare Parts Unavailability.
- 082324 – ASSY,HEAD,PRIMARY,2MM,ICS4 for ICS-4000 systems
- 082602 – HEAD,PRIMARY,PUMP,CAP,ICS5+,CLEANED for capillary pumps. This is just the pump head body; the entire pump head assembly does not exist as a p/n.
- 083477 – HEAD ASSY,PRIMARY,PMP,ICS5+ for Integrion, ICS-5000+/ICS-6000 analytical pumps.
Secondary pump heads:
- 067703 – ASSY, HEAD, SECONDARY, ICS900 for Easion.
- 055733 – HEAD ASSY, SECONDARY, SERIES PK for Aquion, ICS-1X00, ICS-2X00.
- 062054 – HEAD ASSY,SECONDARY,PMP,ICS-3000 for ICS-3000/5000 analytical pumps.
- 082325 – ASSY,HEAD,SECONDARY,2MM,ICS4 for ICS-4000 systems
- 075458 – ASSY,HEAD,SEC,HI PRES UPGRD,TESTED for capillary pumps.
- 083482 – HEAD ASSY,SECONDARY,PMP,ICS5+ for Integrion, ICS-5000+, ICS-6000 analytical pumps.