PSB-387R - IC Validation Test Box
A new IC Validation/Test Box (060615) replaces the EG40 Validation Test Cell (054970) for devices RFC-10, RFC-30, EG40, EG50, ICS-1000, ICS-1500, and ICS-2000. It supports various tests (OQ/PQ, internal/external load tests) and requires an adapter for EG40. It includes leads for connection to ammeter/multimeter.
Description:
A new Validation Test Box 060615 is available for the RFC-10, RFC-30, EG40, EG50, ICS-1000, ICS-1500, and ICS-2000 devices. This test box replaces the EG40 Validation Test Cell 054970.
Environment:
- RFC-10
- RFC-30
- EG40
- EG50
- ICS-1000
- ICS-1500
- ICS-2000
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-387R - IC Validation-Test Box.PDF | January 06, 2022 |