PSB-387 - IC Validation Test Box
A new IC Validation/Test Box (060615) is available for IC systems, replacing the EG40 Validation Test Cell (054970). It supports various tests including OQ/PQ for EG40 and EG50, and RFC-30 power supply tests for SRS, AES, and CR-TC. The box comes with leads for connecting to an ammeter or multimeter, and an adapter cable (057899) for EG40 validation. The calibration is valid for two years. Detailed test procedures for internal and external load tests are provided.
Description:
A new Validation Test Box 060615 is available for IC systems. This test box replaces the EG40 Validation Test Cell 054970.
Environment:
- IC Validation-Test Box 060615
- All SN
- EG50
- RFC-30
- RFC-10
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-387 - IC Validation-Test Box.PDF | January 07, 2022 |