PSB-IC-SYS-1040 - Motor-Position-Sensor-NOV2013
Description:
The “Camshaft Sensor Always Alight” error can be resolved by replacing a sensor on the pump block. Replacing the sensor resolves this error, but does not resolve any other problems or errors.
Environment:
- ICS-5000
- ICS-5000 DP
- ICS-5000 Plus
- ICS-5000 Plus DP
- ICS-6000
- ICS-6000 DP
- Integrion
Attachment(s):
| File | Last Modified |
|---|---|
| psb-ic-sys-1040-motor_position_sensor-nov2013.pdf | March 23, 2022 |