PSB-IC-SYS-1047R - ICS-3000, ICS-5000, ICS-5000 Plus DC module
Description:
When any ICS-3000-5000-5000+ DC module, with SN prior to 14111421, is connected with certain PCs manufactured in 2014 and later including Dell Optiplex 9020, the DC can lose communication with Chromeleon when the DC upper compartment door is closed and temperature is on.
Environment:
- ICS-3000
- ICS-5000
- ICS-5000+
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-IC-SYS-1047R-ICS - ICS-3000, ICS-5000, ICS-5000+ DC module | December 30, 2021 |