PSB-ICSP-1136-Rev B-Integrion Tablet Discontinuation
Description:
Due to an inability to continue sourcing new parts, the Integrion tablets P/N 22153-62200 (KIT,TABLET,INTERNATIONAL,ITGN) and P/N 22153-62100 (KIT,TABLET,NORTH AMERICA,ITGN) have been discontinued. Both parts have been removed from the product guide and a block placed on new orders.
Environment:
Integrion HPIC System with tablet
Recommendations:
1. For customers who desire/require tablet control of their Integrion, it will be necessary for them to purchase a tablet meeting the specifications listed below from an alternate vendor.
2. The Integrion mobile application can be downloaded from the Google Play Store link:
https://play.google.com/store/apps/details?id=com.thermofisher.mobile.android.integrion&hl=en&gl=US
3. The specifications needed for the tablet to function as expected are:
· Screen Size: 7-8”
· Physical size: Shortest side maximum 140mm
· OS: Android
· Minimum Processor/Speed: Quad Core 1.2 GHz
· Minimum Resolution: 1280 X 800
· Minimum Program RAM: 1 GB
· Minimum Storage Memory: 8 GB
· Port: Micro USB or Type-C
4. If a previously installed tablet is covered by a service contract and needs to be replaced, please contact the IC/SP Ion Chromatography Product Support group (ICSP.ProductSupport@thermofisher.com).
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1136-Integrion Tablet Discontinuation Rev B.pdf | June 19, 2026 |