AS-DV misalignments
Issue
AS-DV misalignments
Environment
AS-DV Autosampler
Resolution
Step 1: Press "Carousel Release" button and wait until sample tip is fully up, Turn power off.
Step 2: Remove Motor Assembly and Carousel Assembly plate; see details in AS-DV service manual section 4.14.1 step 1 to 10.
Step 3: Motor and Carousel assembly plate are equipped with two pulleys. Both pulleys are secured by two grub screws. Retighten these 4 screws and use Loctite 221 or similar to secure them again.
Step 4: If necessary replace belt too, P/N 053913.
Step 5: Check for Belt tension; see section 4.14.2 in AS-DV service manual.
Step 6: Reinstall needle assembly and test the alignment performing the needle alignment procedure in section 4.13.1 of the AS-DV service manual.
Step 7: Reinstall tub and carousel.
Step 8: Turn power on, wait for self test, reconnect all modules to Chromeleon.
Step 9: Check for mechanical play of carousel and make at least 3 injections to prove your work.
Cause
AS-DV misalignments are caused by loose set screws.