Andromeda error when configuring suppressor
Issue
-
Andromeda error received when installing or configuring a suppressor
-
Suppressor andromeda configuration error
Environment
-
ICS-5000+
-
ICS-6000
-
Integrion HPIC
-
Aquion IC
-
ICS-1100/2000/2100
Resolution
-
Remove suppressor from configuration
-
Save configuration
-
Stop then start instrument controller
-
Add suppressor back to instrument configuration
-
Select suppressor type inside of Chromeleon console
-
Confirm unit is working as intended