Thermo Fisher Scientific
Collecting Inuvion Analog CD Signal
Issue
Customer would like to use an analog-to-digital box (A2D box, i.e. SatIN, etc) to collect the CD signal.
Environment
 Inuvion
Resolution
The A2D box  will have a cable to connect to the 12-pin green terminal block/connector strip on the back of the Inuvion. Two of the wires will be connected to the two analog pins and two of the wires will be connected to the relay pins. The relay pins are used to trigger the start of data analysis when the injection occurs. Please see Appendix B of the Inuvion Operator Manual.
An example:
processed-932D2E53-A975-4B3C-8FB9-7AD207BC0207.jpeg
In the Chromeleon Instrument Configuration:
NOTE: The Zero Flow Indicator box should be UNCHECKED.
clipboard_ee30f2613e2d8645495c804a1baa35fa3.png
clipboard_e749a334d8a3f519de24e239a0480142e.png
A command will need to be entered into the Instrument method to close the relay in order to start the data collection when the injection occurs.
clipboard_e399e997f65175c7f8584d790b1640eee.png