Thermo Fisher Scientific
Dionex Trap Column Bed Support Part Numbers
Issue
Lookup table for bed suppport part numbers for trap columns.
Environment
Ion Chromatography systems and modules 
Resolution
Part Number
Trap column
BED Support
060477
CR-ATC Column
042955
072078
Dionex CR-ATC Capillary Trap Column
042955
075550
Dionex CR-ATC 500 Trap Column
042955
088662
Dionex CR-ATC 600 Trap Column
042955
079932
Dionex IonPac ATC-3 (2mm) Trap (4x35 mm)
042955
059660
Dionex IonPac ATC-3 (4mm) Trap (9x24 mm)
060270 
079018
Dionex IonPac ATC 500 (2mm) Trap 4x35mm
042955 
075976
Dionex IonPac ATC 500 (4mm) Trap 9x24mm
060270 
059604
Dionex IonPac ATC-HC Anion Trap (9x75mm)
048238 
075978
Dionex IonPac ATC-HC 500 Trap (9x75 mm)
048238 
066262
PROD,CONS,CR-CTC II
042955
104-60001
PROD,COL,CR-CTC III,TRACKING ENABLED
042955
072079
Dionex CR-CTC Capillary Trap Column
042955
075551
Dionex CR-CTC 500 Trap Column
042955
088663
PROD,COL,CR-CTC600,TRACKING ENABLED
042955
043132
Dionex IonPac CTC (2mm) Trap (4x35mm)
042955 
040192
Dionex IonPac CTC-1 (4mm) Trap (9x24mm)
039037 
079019
Dionex IonPac CTC 500 (2mm) Trap 4x35mm
042955 
075977
Dionex IonPac CTC 500 (4mm) Trap 9x24mm
042955 
  • Was this article helpful?