High-Performance Anion Exchange - Pulsed Amperometric Detection
The attached describes HPAE-PAD, which refers to a technique where non-derivatized carbohydrates (ranging from simple sugars to complex carbohydrates) are analyzed by High-Performance Anion-Exchange (HPAE) chromatography coupled with Pulsed Amperometric Detection (PAD). The document also describes dual eluent generation for carbohydrate analysis.
Description:
This document contains information on the HPAE-PAD for Carbohydrate analysis. HPAE-PAD refers to a technique where non-derivatized carbohydrates (ranging from simple sugars to complex carbohydrates) are analyzed by High-Performance Anion Exchange (HPAE) chromatography coupled with Pulsed Amperometric Detection (PAD).
Environment:
Ion Chromatography
Attachment(s):
| File | Last Modified |
|---|---|
| A New, Simple and Automated Workflow Solution for Carbohydrate Analysis.pdf | December 30, 2021 |