High pressure then no pressure
Issue
-
Bottom pump behaving erratically
-
High pressure then no pressure
-
Pressure typically found at 2000 psi, now at 2800 psi and overpressured
Environment
- ICS6000 DP Bottom Pump
Resolution
-
Attempt power cycle.
-
Attempt to prime pump.
-
If issue is unresolved, replace guard and column.
-
Turn pump off and wait a full 60 seconds before restarting pump. Pressure issue should be resolved.